Recommended Products
L
19 cm
Quality Level
input
sample type pharmaceutical(s)
sample type beer
sample type cosmetics
sample type bottled water
application(s)
bioburden testing
cosmetics
environmental
food and beverages
water monitoring
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service