This product is compatible with swinging-bucket capable of handling 250 mL bottles. Do not use a fixed angle rotor. Kindly review more technical specifications in the User Guide under "More Documents" or at this link: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/866/336/pr03221w-ms.pdf
UFC710008
Centricon® Plus Centrifugal Filter
MWCO 100 kDa, sample volume 15-70 mL, regenerated cellulose membrane
Synonym(s):
Centrifuge concentrator
About This Item
Recommended Products
material
polypropylene cap
polypropylene cup (for concentrate/retentate)
polypropylene cup (for filtrate collection)
polypropylene cup (for sample filter)
regenerated cellulose membrane
Quality Level
sterility
non-sterile
product line
Amicon®
feature
holdup volume 75 μL
manufacturer/tradename
Amicon® Ultra
Centricon®
parameter
15-70 mL sample volume
technique(s)
protein extraction: suitable
protein purification: suitable
L
121 mm
diam.
6 cm
filtration area
19 cm2
working volume
15—70 mL
pore size
100 kDa MWCO
application(s)
sample preparation
shipped in
ambient
General description
The Centricon Plus-70 centrifugal filter is a disposable, single-use device designed for the processing of aqueous biological solutions in volumes ranging from 15 to 70 mL.
Application
Features and Benefits
- compatiblewith swinging-bucket centrifuges only
- can concentrate most 70 mL solutionsdown to 350 µL in 15-40 minutes
Physical form
Other Notes
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Was für ein Rotor wird für die Filter benötigt? What kind of rotor is required for the filter units?
1 answer-
Helpful?
-
-
What are the proposed adapters for centrifugation? We have a swing-bucket centrifuge in the lab, but we don´t have the proper bucket for it.
1 answer-
This device is suitable for swinging bucket rotors only. These centrifugal filters measure 121 mm (length) x 60 mm (diameter) and have a maximum g-force of 3500 x g. See the link below to review the recommended rotors and adaptor for several major brands of centrifuge:
https://www.emdmillipore.com/life-science-research/protein-sample-preparation/rotor-guides/Centricon-Plus-70/2B2b.qB.isYAAAFEcTQMfo3B,navHelpful?
-
-
What is the maximum this device can be spun without damaging the membrane?
1 answer-
The maximum centrifugal force for concentrating is 3,500 × g. For more details please refer to the link below: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/866/336/pr03221w-ms.pdf
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service