Skip to Content
Merck
All Photos(1)

Documents

TX0280

Supelco

Tetrahydrofuran

≥99.5% (GC), Meets ACS Specifications, General Reagents grade, contains 0.0140-0.0310% BHT as stabilizer

Synonym(s):

Diethylene oxide, THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-726-8

product name

Tetrahydrofuran, Meets ACS Specifications GR ACS

vapor density

2.5 (vs air)

Quality Level

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

610 °F

contains

0.0140-0.0310% BHT as stabilizer

expl. lim.

1.8-11.8 %

impurities

≤0.005% Peroxide (as H2O2)
≤0.05% Water (H2O)

evapn. residue

≤0.03%

color

APHA: ≤10

refractive index

n20/D 1.407 (lit.)

pH

7-8 (20 °C, 200 g/L in H2O)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

transition temp

flash point -21.5 °C

density

0.889 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications GR ACS
TX0280P-1 is packaged in poly coated glass bottles

Analysis Note

Assay (GC): 99.5% min
Color (APHA): 10 max
Evaporation residue: 0.03% max
Form: Clear liquid
Peroxide (as H2O2): 0.005% max
Stabilizer (BHT): 0.0140-0.0310%
Water (H2O): 0.05% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service