Skip to Content
Merck
All Photos(2)

Key Documents

SMWP02500

Millipore

MCE Membrane Filter, 5.0 μm Pore Size

greener alternative

MF-Millipore, filter diam. 25 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 5 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

Agency

in accordance with ASTM® D5755

description

25 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

sustainability

Greener Alternative Product

parameter

32 L/min-cm2 air flow rate
580 mL/min-cm2 water flow rate
75 °C max. temp.

diam.

25 mm

filter L × W

1.9 cm × 4.2 cm

filter diam.

25 mm

thickness

135 μm

gravimetric extractables

6%

refractive index

n/D 1.495

matrix

MF-Millipore

pore size

5.0 μm pore size
84 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥0.42 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: SMWP
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • QC of fluid holding tanks
  • Fluid monitoring
  • Particle collection
  • Particle analysis

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service