Skip to Content
Merck
All Photos(1)

Key Documents

SCT244

Sigma-Aldrich

BioTracker Dual-Color Mitochondria-Lysosome Live Cell Dye

Synonym(s):

Live Cell Imaging Probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.21

mol wt

calculated mol wt 586.46 kDa

Quality Level

packaging

vial of 1 mg

manufacturer/tradename

Millipore

storage condition

protect from light

growth mode

N/A

technique(s)

cell based assay: suitable
cell culture | stem cell: suitable
flow cytometry: suitable

detection method

fluorometric

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

General description

The BioTracker Dual-Color Mitochondria-Lysosome Live Cell Dye (Coupa) is a fluorescent probe used to detect mitochondria-lysosome interactions (Chen, 2020).


Mitochondrial-lysosomal interactions are fundamental to the processes of cellular homeostasis. Mitochondria and lysosomes have unique interactions which involve transfer of metabolites. Some disease pathogenesis can be linked to this important subcellular interaction (Chen, 2020). This BioTracker live cell probe is a unique cell-permeable and viscosity-responsive molecular probe that is capable of monitoring mitochondrial and lysosomal interactions in living cells. Through fluorescence conversion, this BioTracker dual color probe can simultaneously tag mitochondria with blue fluorescence and lysosomes with red fluorescence.


Spectral Properties


Fluorescence images obtained by λex = 405 nm and emission at 450 nm (Mitochondria) and λex = 561nm and emission at 590nm (Lysosome)


Reference
Chen, Qixin, et al. A dual-labeling probe to track functional mitochondria–lysosome interactions in live cells." Nature Communications 11.1 (2020): 1-10."

Application

Purity > 98% confirmed by HPLC. Identification confirmed by HNMR, LC-MS, and elemental analysis.

Features and Benefits

Dual color (blue and red) small molecule probe for visualizing mitochondria-lysosome interactions during mitophagy.

Physical form

Lyophilized. Brown solid.

Storage and Stability

Store at -20°C, dessicated and protected from light. Note: Centrifuge vial briefly to collect contents at bottom of vial before opening

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service