Skip to Content
Merck
All Photos(1)

Key Documents

SCC152

Sigma-Aldrich

1HAEo- Human Airway Epithelial Cell Line

Human

Synonym(s):

1HAEo

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

1HAEo- Human Airway Epithelial Cell Line, 1HAEo- human airway epithelial cell line is a useful model for the study of epithelial ion transport, secretion and biochemistry.

biological source

human

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable

shipped in

ambient

General description

1HAEo- is a human airway epithelial cell line that was immortalized with the origin-of-replication defective SV40 plasmid (pSVori-). The cell line retains characteristic features of normal differentiated airway epithelial cells including a cobblestone morphology, cytokeratin expression, the ability to form tight junctions and directional ion transport. It is unknown whether the donor had cystic fibrosis or not, however the cells do not contain the ΔF508 mutation in the CFTR gene or other CFTR mutations presenting 98% of all CF mutations (1). CFTR sequence analysis revealed only wild-type CFTR sequences, supporting the conclusion that 1HAEo- are derived from normal (i.e. non-cystic fibrosis) tissue (1).

Cell Line Description

Epithelial Cells

Application

1HAEo- human airway epithelial cell line is a useful model for the study of epithelial ion transport, secretion and biochemistry.
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X10⁶ viable cells.
• Cells are tested by PCR and are negative for HPV-16, HPV-18, Hepatitis A, C, and HIV-1 & 2 viruses as assessed by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Cozens et al.
In vitro cellular & developmental biology : journal of the Tissue Culture Association, 28A(11-12), 735-744 (1992-11-01)
The successful establishment of a postcrisis SV-40 T antigen transformed epithelial cell line, 1HAEo-, which retains tight junctions and vectorial ion transport, is described. Immunocytochemical analysis of 1HAEo- cells shows a defined pattern of cytokeratin staining and a characteristic pericellular
Sarmad Al-Sahaf et al.
International journal of cancer, 144(2), 334-344 (2018-09-08)
Human papillomavirus (HPV) is now recognised as a major aetiological agent in the pathogenesis of oropharyngeal carcinoma (OPC). HPV-positive tumours are associated with better outcomes compared to HPV-negative tumours, possibly due to differences in their aetiology and/or the tumour microenvironment.
Alexandra Ruth Glathar et al.
Frontiers in oncology, 12, 879054-879054 (2022-06-18)
The complex heterogeneity of head and neck squamous cell carcinoma (HNSCC) reflects a diverse underlying etiology. This heterogeneity is also apparent within Human Papillomavirus-positive (HPV+) HNSCC subtypes, which have distinct gene expression profiles and patient outcomes. One aggressive HPV+ HNSCC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service