Skip to Content
Merck
All Photos(1)

Key Documents

SCC058

Sigma-Aldrich

FibroGRO® Xeno-Free Human Foreskin Fibroblasts

The FibroGRO Xeno-Free Human Foreskin Fibroblasts are derived from normal human foreskin & have been isolated & manufactured under xeno-free conditions.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.75

biological source

human foreskin (fibroblasts)

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Chemicon®
FibroGRO®
Millipore

morphology

(endothelial)

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

shipped in

liquid nitrogen

General description

Product Source: Human foreskin fibroblasts, derived under xeno-free conditions

Cell Line Origin

Skin

Cell Line Description

Fibroblast Cells

Application

Research Category
Stem Cell Research

Components

1 vial of Human Foreskin Fibroblasts

Quality

Negative for mycoplasma, HTLV-1&2, Hepatitis B&C, HIV, Treponema pallidum.

Storage and Stability

Store in Liquid Nitrogen

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
FIBROGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service