Skip to Content
Merck
All Photos(1)

Documents

SCC043

Sigma-Aldrich

C28/I2 Human Chondrocyte Cell Line

Human

Synonym(s):

C-28/I2, C28/I2, Human chondrocyte cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

C28/I2 Human Chondrocyte Cell Line, C28/I2 Human Chondrocyte Cell Line is widely used as a model cell line for studying normal and pathological cartilage repair mechanisms related to chondrocyte biology and physiology.

biological source

human

Quality Level

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

General description

The chondrocyte is the only specialized cell type present in articular cartilage. They produce secretions to support and repair the cartilage matrix. Human chondrocytes in mature articular cartilage are post-mitotic and terminally differentiated cells. Studies on human chondrocytes have been hampered by the difficulty in obtaining sufficient numbers of primary human chondrocytes from a single joint and by variabilities among donors due to factors such as age and medical conditions.

C-28/I2 is a clonal line derived from the nonclonal T/C-28a4 cells which were immortalized via retroviral vector-mediated simian virus SV40 Large T antigen (Tag) expression.

Cell Line Description

Chondrocyte Cells

Application

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested by PCR and are negative for HPV-16, HPV-18, Hepatitis A, B, C and HIV-1 & 2 viruses.
• Cells are negative for mycoplasma contamination.
• Each lot of cells are genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Libo Zhou et al.
ACS applied materials & interfaces, 13(43), 51321-51332 (2021-10-20)
Various biomaterial scaffolds have been developed for improving stem cell anchorage and function in tissue constructs for in vitro and in vivo uses. Growth factors are typically applied to scaffolds to mediate cell differentiation. Conventionally, growth factors are not strictly
Qunli Wang et al.
Bioengineered, 12(2), 12812-12820 (2021-12-14)
Osteoarthritis (OA) is a cartilage degenerative disease commonly observed in the elderly population and significantly impacts the normal life of OA patients. It has been reported that the development of pathological cell senescence in chondrocytes is involved in the pathogenesis
Meng-Huang Wu et al.
International journal of molecular sciences, 20(1) (2018-12-28)
Chondrosarcoma, a heterogeneous malignant bone tumor, commonly produces cartilage matrix, which generally has no response to conventional therapies. Studies have reported that MLN4924, a NEDD8-activating enzyme inhibitor, achieves antitumor effects against numerous malignancies. In this study, the suppressive effects of
Yanting Rao et al.
Immunity, inflammation and disease, 11(10), e978-e978 (2023-10-31)
Rheumatoid arthritis (RA) is a chronic inflammation mediated by an autoimmune response. Baculoviral IAP repeat-containing 2 (BIRC2) and tumor necrosis factor receptor 1-associated death domain protein (TRADD) have been reported to be highly expressed in RA, while their specific roles
Dong-Yeong Lee et al.
Antioxidants (Basel, Switzerland), 9(1) (2020-01-23)
Antioxidant enzymes are decreased in osteoarthritis (OA) patients, implying the role of oxidative stress in osteoarthritis pathogenesis. The aim of this study was to evaluate the cytoprotective effects of delphinidin, a potent antioxidant, in human chondrocytes and the underlying mechanisms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service