PHCC340500
Scepter 3.0 Cell Counter Sensors
40 μm sensors designed to count particles between 4 μm & 18 μm
Synonym(s):
Scepter sensors
About This Item
Recommended Products
feature
50 μL (sample volumn)
packaging
pkg of 500 ea
manufacturer/tradename
Scepter
parameter
50000 particles/mL particle concentration (1500000 particles/mL)
technique(s)
cell counting: suitable
particle size
4-18 μm
compatibility
Scepter for use with cells sized 5.0–15.0 μm
Not compatible with Scepter 2.0 Instruments
shipped in
ambient
General description
The Scepter 3.0 cell counter′s screen displays:
- Cell concentration
- Average cell size
- Average cell volume
- Histogram of size or volume distribution
This system is intended for research use only and has been tested with cell types representative of those in use today.
Features and Benefits
Coupled with precision liquid handling channels and electronics, the Scepter sensor accurately and reliably provides cell population statistics
Other Notes
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service