Skip to Content
Merck
All Photos(4)

Key Documents

MZHABG251

Millipore

Microfil® with EZ-Pak®

pore size 0.45 μm

Synonym(s):

Membrane filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Microfil® with EZ-Pak® is an easy to use, fast and accurate system for the routine microbiological analysis of beverages and drinking water. For the ultimate convenience and efficiency, combine the EZ-Pak® system with the Microfil® filtration unit. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use 250 mL Microfil® funnels that comes with the 0.45 μm pore size, black gridded EZ-Pak® filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and more efficient to trap bacteria. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in consistent recovery of organisms.

Application

Microfil® with EZ-Pak® filters find application in Clinical, Cosmetics, Environmental, Industrial QC, Pharmaceutical, Food, and Beverage industries. Microbial analysis of beer, bottled water, cider, sports drink, and wine by membrane filtration method

Features and Benefits

  • Convenient and easy to use
  • Sterilized and ready-to-use device - saves time and expense
  • Reduce laboratory workload
  • Faster filtration rate due to larger surface area

Principle

Vacuum

Physical form

Color Code: Black

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service