Skip to Content
Merck
All Photos(2)

Key Documents

MSCRMAG-42K

Millipore

MILLIPLEX® Mouse Soluble Cytokine Receptor Magnetic Bead Panel - Immunology Multiplex Assay

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 89-98%
standard curve range: 12.2-50,000 pg/mL
(sIL-1RI, sIL-1RII, sIL-2Rα, sIL-4R, sTNFRII)

standard curve range: 24.4-100,000 pg/mL
(sCD30, sgp130, sIL-6R, sRAGE, sVEGFR1, sVEGFR2, sVEGFR3)

standard curve range: 4.9-20,000 pg/mL
(sTNFRI)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Cytokine receptors constitute an integral part of the cytokine biology. Like cytokines, cytokine receptors are involved in normal physiological and pathological processes of almost all disease states. Soluble cytokine receptors naturally arise from genes encoding membrane-bound receptors or are direct derivatives of the receptors themselves. The discovery that soluble cytokine receptors are involved in regulating excessive inflammatory responses and modulating immune events has stimulated significant research interest in their potential role as immunotherapeutic agents. Many of these soluble cytokine receptors have the ability to inhibit the binding and biological activity of their cytokine ligands, making them very specific cytokine antagonists.

The MILLIPLEX® Mouse Soluble Cytokine Receptor Panel contains 13 metastasis biomarkers that have also been identified in a variety of therapeutic areas. Coupled with the Luminex® xMAP® platform in bead format, this panel enables researchers to focus on discovery with the advantage of simultaneous quantitative multiplex detection leading to speed and sensitivity.

Panel Type: Cytokines/Chemokines

Application

  • Analytes: sCD30 (sTNFRSF8), sgp130, sIL-1RI (sCD121a), sIL-1RII (sCD121b), sIL-2Rα (sCD25), sIL-4R (sCD124), sIL-6R (sCD126), sRAGE, sTNFRI (sTNFRSF1A), sTNFRII (sTNFRSF1B), sVEGFR1 (sFlt-1), sVEGFR2 (sFlk-1), sVEGFR3 (sFlt-4)
  • Recommended Sample type: serum, plasma and cell culture
  • Recommended Sample dilution: 1:5 diluted serum and plasma
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saida Breuer et al.
Nutrients, 13(11) (2021-11-28)
Maternal obesity greatly affects next generations, elevating obesity risk in the offspring through perinatal programming and flawed maternal and newborn nutrition. The exact underlying mechanisms are poorly understood. Interleukin-6 (IL-6) mediates its effects through a membrane-bound receptor or by trans-signaling
Damien Restagno et al.
PloS one, 11(8), e0162109-e0162109 (2016-08-31)
Sepsis is characterized by pro- and anti-inflammatory responses following infection. While inflammation is responsible for widespread organ damage, anti-inflammatory mediators lead to immunoparalysis increasing susceptibility to secondary infections (nosocomial pneumonia). We aimed to investigate the impact of bacterial load on
Susana Rodriguez et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 320(1), R19-R35 (2020-10-22)
C1q/TNF-related protein 1 (CTRP1) is an endocrine factor with metabolic, cardiovascular, and renal functions. We previously showed that aged Ctrp1-knockout (KO) mice fed a control low-fat diet develop renal hypertrophy and dysfunction. Since aging and obesity adversely affect various organ
Katherine Young Bezold Lamm et al.
eLife, 7 (2018-01-09)
Healthy pregnancy depends on proper placentation-including proliferation, differentiation, and invasion of trophoblast cells-which, if impaired, causes placental ischemia resulting in intrauterine growth restriction and preeclampsia. Mechanisms regulating trophoblast invasion, however, are unknown. We report that reduction of Inverted formin 2
Susana Rodriguez et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 2657-2676 (2020-01-08)
Local and systemic factors that influence renal structure and function in aging are not well understood. The secretory protein C1q/TNF-related protein 1 (CTRP1) regulates systemic metabolism and cardiovascular function. We provide evidence here that CTRP1 also modulates renal physiology in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service