MR-106
EmbryoMax® KSOM Mouse Embryo Media
(1X), Liquid, with 1/2 Amino Acids
Synonym(s):
K+ Simplex Optimised Medium (KSOM), KSOM Mouse Embryo Media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Specialty Media
EmbryoMax®
technique(s)
cell culture | embryo: suitable
cell culture | stem cell: suitable
input
sample type: mouse embryo(s)
Application
EmbryoMax® KSOM Mouse Embryo Media has been used in the culturing of mouse embryos for embryonic development.
Storage and Stability
Can be stored at -20°C upto the expiration date on the bottle.
Once thawed it should be used within 2 weeks.
Note: Our EmbryoMax Liquid Mouse Embryo Media is produced on a bi-monthly basis. For regular users, we encourage standing orders to ensure that backorders do not occur.
Once thawed it should be used within 2 weeks.
Note: Our EmbryoMax Liquid Mouse Embryo Media is produced on a bi-monthly basis. For regular users, we encourage standing orders to ensure that backorders do not occur.
Legal Information
EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Benjamin L Kidder
Methods in molecular biology (Clifton, N.J.), 1150, 191-199 (2014-04-20)
Epigenetic regulation of gene expression in the germline is important for reproductive success of mammals. Misregulation of genes whose expression is correlated with reproductive success may result in subfertility or infertility. To study epigenetic events that occur during oocyte maturation
Yilong Miao et al.
STAR protocols, 2(1), 100298-100298 (2021-02-04)
The low quality of oocytes is one of the main causes of the suboptimal reproductive outcome of female mammals with advanced maternal age. Here, we present a detailed protocol to obtain high-quality oocytes and embryos from aged mice by nicotinamide
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service