Skip to Content
Merck
All Photos(1)

Key Documents

MPTA

Millipore

Adapters for mPAGE® gels when using XCell SureLock® and other compatible electrophoresis tanks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NA.56

grade

electrophoresis grade

packaging

pack of 2 ea

General description

Adapters for mPAGE gels when using XCell SureLockand other compatible electrophoresis tanks.
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

Use these adapters with XCell SureLock and other compatible electrophoresis tanks. Not required when using Bio-Rad Mini-PROTEAN systems.

  • Western Blotting

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany
XCell SureLock is a registered trademark of Life Technologies

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service