Skip to Content
Merck
All Photos(1)

Key Documents

MP81G15

Millipore

mPAGE® 8-16% Bis-Tris Precast Gels

8-16% Bis-Tris, 15 well, 1mm

Synonym(s):

Bis-Tris gel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105320
NACRES:
NC.01

product name

mPAGE® 8-16% Bis-Tris Precast Gel, L × W × wells 10 cm × 8 cm × 15 well

shelf life

18 mo.

packaging

pkg of 10 gels

L × W × wells

10 cm × 8 cm × 15 well

cassette W × L

10 cm × 8 cm

gel thickness

1 mm

well maximum volume

40 μL

well wells

15 wells

compatibility

mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

mPAGE 8-16% Bis-Tris Precast Gel, 10x8 cm
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

mPAGE 8-16% bis-tris gels for high resolution protein separation
  • Load up to 60 μl sample / well
  • Typical run time 26 min (MES, 180V) or 31 min (MOPS, 200V)
  • Reverse gasket when using with Bio-Rad Mini-PROTEAN systems
  • Use adapter (MPTA) with XCell SureLock and other compatible systems


Western Blotting

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service