MP41G12TR2
mPAGE® Trial Kit
includes two 4-12% Bis-Tris precast gels, 10x8 cm, 12-well, MES SDS powder, makes 1L of running buffer, 2 tank adapters
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105300
NACRES:
NA.56
Recommended Products
feature
includes two 4-12% Bis-Tris precast gels, 10x8 cm, 12-well, MES SDS powder, makes 1L of running buffer, 2 tank adapters
Looking for similar products? Visit Product Comparison Guide
General description
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Application
mPAGE 4-12% bis-tris gels for high resolution protein separation
Western blotting
- Load up to 60 μl sample / well
- Typical run time 26 min (MES, 180V) or 36 min (MOPS, 200V)
- Reverse gasket when using with Bio-Rad Mini-PROTEAN systems
- Use adapter (MPTA) with XCell SureLock and other compatible systems
Western blotting
Legal Information
MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service