MMHVWP124
Milliflex Oasis® Filtration Unit
pore size 0.45 μm, PVDF membrane, white filter, volume 100 mL
Synonym(s):
Sterile membrane filtration device for bioburden testing
About This Item
Recommended Products
material
PVDF membrane
plain filter
styrene-butadiene device
white filter
Quality Level
Agency
EP, USP, JP
sterility
electron beam irradiated
feature
ready-to-use
packaging
pack of 24
manufacturer/tradename
Milliflex Oasis®
capacity
100 mL
filter diam.
49 mm
funnel capacity
100 mL
volume
100 mL
pore size
0.45 μm pore size
application(s)
bioburden testing
cosmetics
pharmaceutical
water monitoring
compatibility
for use with Milliflex Oasis® System
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The PVDF membrane filter has a broad chemical compatibility and low protein binding.
The MILLIFLEX® PROTACT feature protects your samples from secondary contamination and makes the membrane-to-agar contact optimal delivering the most reliable results. The patented sealed membrane filter feature prevents the bypass phenomenon often found with other filtration systems ensures robust read-out.
The MILLIFLEX® AZURE feature combines transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative filtration unit assembly design prevents any leaks or sample by-pass of the membrane to ensures the right sample volume is filtered.
The MILLIFLEX® RACK&STACK feature also includes a locked lid for safe transportation and incubation.
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service