Skip to Content
Merck
All Photos(1)

Documents

MABT188

Sigma-Aldrich

Anti-Dynamin-1/2 Antibody, clone Hudy-1

clone Hudy-1, from mouse

Synonym(s):

Dynamin-1, Dynamin, Dynamin-1/2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Hudy-1, monoclonal

species reactivity

human, rat

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DNM1(1759)

General description

Dynamin-1 & Dynamin-2 belong to the dynamin superfamily of large GTPases. Dynamins are multidomain proteins consisting of a GTP-binding domain, a middle domain, and a GTP-effector domain. Dynamins also contain a pleckstrin homology (PH) domain which may facilitate membrane binding. Dynamin-1 is highly abundant in the brain particularly at synaptic contacts, whereas Dynamin-2 is ubiquitously expressed in non-neuronal cells. Dynamins are involved in receptor-mediated endocytosis of clathrin-coated vesicles. Dynamin-1 interacts with an array of cellular targets via its SH3 binding domain and is implicated in late stages of vesicle formation by forming oligomers around the “neck” of budding vesicles and inducing membrane fission; it may also play a role in cytoskeleton rearrangement and cytoskinesis. Dynamin-2 is involved in the formation of clathrin-coated vesicles but may also be involved in the recycling of receptors between endosomes and the trans-Golgi network.

Specificity

This antibody detects both Dynamin-1 and Dynamin-2. (Nicoziani, P., et al. (1999). Molecular Biology of the Cell. 11:481–495.)

Immunogen

Recombinant full length human Dynamin-1 expressed in transfected Sf9 cells.

Application

Anti-Dynamin-1/2 Antibody, clone Hudy-1 detects level of Dynamin-1/2 & has been published & validated for use in Western Blotting, Immunofluorescence.
Immunofluorescence Analysis: A representative lot was used by an independent laboratory in HeLa cells (Nicoziani, P., et al. (1999). Molecular Biology of the Cell. 11:481–495.)
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Dynamin-1/2 on 10 µg of rat brain tissue lysate.

Target description

~97 kDa observed.
Dynamin 1 has a calculated MW of 97 kDa Dynamin 2 has a calcluated MW of 98 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4) and 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamalika Mukherjee et al.
Nature communications, 13(1), 2422-2422 (2022-05-04)
Chronic kidney diseases and acute kidney injury are mechanistically distinct kidney diseases. While chronic kidney diseases are associated with podocyte injury, acute kidney injury affects renal tubular epithelial cells. Despite these differences, a cardinal feature of both acute and chronic
Hyun Jung Park et al.
Frontiers in physiology, 11, 587358-587358 (2020-11-17)
Cultured cell models are an essential complement to dissecting kidney proximal tubule (PT) function in health and disease but do not fully recapitulate key features of this nephron segment. We recently determined that culture of opossum kidney (OK) cells under
Cinzia Milesi et al.
Life science alliance, 2(1) (2019-01-30)
EPS15 and its homologous EPS15L1 are endocytic accessory proteins. Studies in mammalian cell lines suggested that EPS15 and EPS15L1 regulate endocytosis in a redundant manner. However, at the organismal level, it is not known to which extent the functions of
Elin Larsson et al.
The Journal of cell biology, 222(4) (2023-02-03)
Caveolae are small membrane invaginations that generally are stably attached to the plasma membrane. Their release is believed to depend on the GTPase dynamin 2 (Dyn2), in analogy with its role in fission of clathrin-coated vesicles. The mechanistic understanding of
Dominik P Buser et al.
EBioMedicine, 46, 32-41 (2019-07-25)
Gliomas are the most frequent and aggressive malignancies of the central nervous system. Decades of molecular analyses have demonstrated that gliomas accumulate genetic alterations that culminate in enhanced activity of receptor tyrosine kinases and downstream mediators. While the genetic alterations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service