Skip to Content
Merck
All Photos(1)

Key Documents

MABT117

Sigma-Aldrich

Anti-Endoglin (CD105) Antibody, clone 44G4

clone 44G4, from mouse

Synonym(s):

Endoglin, CD104

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

44G4, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ENG(2022)

General description

Endoglin (CD105) is a homodimeric integral membrane glycoprotein comprised of a 95 kDa subunit. Endoglin is a component of the TGF-β receptor system that functions as an ancillary receptor influencing binding of the Transforming Growth Factor beta (TGF-β) family of ligands to signaling receptors. Endoglin binds to TGF-β1 and TGF-β3 with high affinity, but does not bind TGF-β2. Endoglin is expressed by vascular endothelial cells and syncytiotrophoblasts of placenta and is weakly expressed by stromalfibroblasts. Endoglin is also expressed by U937 cells and by activated macrophages. Endoglin expression is up-regulated on activated endothelium in tissues undergoing angiogenesis, such as in tumors, or in cases of wound healing or dermal inflammation.

Immunogen

Whole HOON Pre-B Leukemic Cells

Application

Anti-Endoglin (CD105) Antibody, clone 44G4 is an antibody against Endoglin (CD105) for use in FC, IH & IP.
Immunohistochemistry Analysis: A representative lot was used by an independent laboratory to detect Endoglin in normal human lymph node, tonsil, spleen thrymus, liver, breast, lung, and kidney specimens (Gougos, A., and Letarte, M. (1988). J Immunol. 141(6):1925-1933).

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to IP Endoglin from HOON, NALM-6, 3104, 3161, HL-60, U-937, EM-3 and KG-1a cells (Gougos, A., and Letarte, M. (1988). J Immunol. 141(6):1925-1933).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Flow Cytometry in HUVEC cells.

Flow Cytometry Analysis: A 1:200 dilution of this antibody detected Endoglin in HUVEC cells.

Target description

71 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HUVEC cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Can Intranasal Administration of Adipose-Derived Stem Cells Reach and Affect the Histological Structure of Distant Organs of Aged Wistar Rat?
Hamam, et al.
Journal of microscopy and ultrastructure, 11, 11-Jan-11-Jan (2023)
Arantxa Blázquez-Prunera et al.
Stem cell research & therapy, 8(1), 103-103 (2017-04-30)
Mesenchymal stem cells (MSCs) show promising characteristics for their use in advanced therapy medicinal products. However, there are some unresolved concerns, such as the use of animal components for their expansion. In this study we assessed the suitability of a
Qiuyun Li et al.
Molecular medicine reports, 24(3) (2021-07-24)
Hepatic ischemia/reperfusion injury (HIRI) is a complex pathophysiological process that may develop after liver transplantation and resection surgery, as well as in uncontrolled clinical conditions. Bone marrow‑derived mesenchymal stem cells (BM‑MSCs) are potential targets for liver diseases. Thus, the present
José M Díez et al.
Stem cell research & therapy, 6, 28-28 (2015-04-19)
Fetal bovine serum (FBS) is an animal product used as a medium supplement. The animal origin of FBS is a concern if cultured stem cells are to be utilized for human cell therapy. Therefore, a substitute for FBS is desirable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service