Skip to Content
Merck
All Photos(2)

Key Documents

MABS195

Sigma-Aldrich

Anti-Pan-Ras Antibody, clone RAS 10

clone RAS 10, from mouse

Synonym(s):

GTPase Kras, K-Ras 2, Ki-Ras, c-K-ras, c-Ki-ras, GTPase Kras, N-terminally processed

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RAS 10, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KRAS(3845)

General description

The human ras gene family consists of three identified members H-, K- and N-ras encoding proteins of 188-189 amino acids and 21,000 (p21) daltons. Human H- and K-ras are the homologues of v-H- and v-K-ras, sequences originally isolated from Harvey and Kirsten strains of rat sarcoma viruses. Normal human cellular ras genes are referred to as proto-oncogenes, and have the potential for activation to oncogenes by mutations occurring in codons 12, 13 and 61; such mutated, activated and transforming ras genes have been identified and isolated from human tumors and cultured tumor cells. Although the expression patterns of ras proto-oncogene proteins in normal human tissues are known, similar information for activated ras oncogene encoded p21s and their relevance to human disease diagnosis and prognosis is still emerging.

Immunogen

Recombinant protein corresponding to human Pan-Ras.

Application

Detect Pan-Ras using this Anti-Pan-Ras Antibody, clone RAS 10 validated for use in Western Blotting, ICC.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
Western Blot Analysis: A representative lot of this antibody detected Pan-Ras in SW-480 cell lysate.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Pan-Ras in A431 cells.

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Pan-Ras in IP (Hamer, P.J., et al. (1990). Hybridoma. 9(6):573-587.).

Quality

Evaluated by Western Blot in MCF7 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Pan-Ras in 10 µg of MCF7 cell lysate.

Target description

~21 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Won-Ji Ryu et al.
Experimental & molecular medicine, 52(5), 832-842 (2020-05-28)
Triple-negative breast cancer (TNBC) is a severe and heterogeneous disease that lacks an approved targeted therapy and has a poor clinical outcome to chemotherapy. Although the RAS-ERK signaling axis is rarely mutated in TNBC, ~50% of TNBCs show an increased
Alisa Damnernsawad et al.
Haematologica, 107(1), 77-85 (2020-12-31)
Drug resistance impedes the long-term effect of targeted therapies in acute myeloid leukemia (AML), necessitating the identification of mechanisms underlying resistance. Approximately 25% of AML patients carry FLT3 mutations and develop post-treatment insensitivity to FLT3 inhibitors, including sorafenib. Using a
Won-Ji Ryu et al.
Cancers, 11(4) (2019-04-11)
: Treatment of advanced gastric cancer patients with current standard chemotherapeutic agents frequently results in resistance, leading to poor overall survival. However, there has been no success in developing strategies to overcome it. We showed the expression levels of both
Sang-Kyu Lee et al.
EMBO reports, 19(12) (2018-11-11)
RAS proteins play critical roles in various cellular processes, including growth and transformation. RAS proteins are subjected to protein stability regulation via the Wnt/β-catenin pathway, and glycogen synthase kinase 3 beta (GSK3β) is a key player for the phosphorylation-dependent RAS
Michele Caraglia et al.
International journal of cancer, 121(10), 2317-2330 (2007-07-28)
Interferon alpha (IFNalpha) induces an EGF-Ras-->Raf-1-->Erk dependent survival pathway counteracting apoptosis induced by the cytokine. In this paper we have evaluated the effects of the combination between farnesyl-transferase inhibitor (FTI) R115777 and IFNalpha on the growth inhibition and apoptosis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service