Skip to Content
Merck
All Photos(1)

Documents

MABS1233

Sigma-Aldrich

Anti-HMG-CoA Reductase Antibody, clone IgG-A9

clone IgG-A9, from mouse

Synonym(s):

3-hydroxy-3-methylglutaryl-coenzyme A reductase, HMG-CoA reductase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

IgG-A9, monoclonal

species reactivity

human, rat, hamster

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HMGCR(3156)

General description

3-hydroxy-3-methylglutaryl-coenzyme A reductase (EC 1.1.1.34; UniProt P00347; also known as HMG-CoA reductase) is encoded by the HMGCR gene (Gene ID 100756363) in Cricetulus griseus (Chinese hamster). HMG-CoA reductase catalyzes the conversion of HMG-CoA to mevalonic acid, the rate-limiting enzyme of the mevalonate-isoprenoid biosynthesis (MIB) pathway responsible for the production of cholesterol and other isoprenoids, as well as farnesyl pyrophosphate (FPP) required for protein prenylation. HMG-CoA reductase is suppressed by cholesterol derived from the internalization and degradation of low density lipoprotein (LDL) via the LDL receptor as well as oxidized cholesterol species. Competitive inhibitors against HMG-CoA reductase induce LDL receptors expression in the liver, which in turn increases the catabolism of plasma LDL and lowers the plasma concentration of cholesterol. HMG-CoA reductase is a multitransmembrane ER protein with an N-terminal sterol-sensing domain (SSD; a.a. 61-218) and a C-terminal catalytic domain (a.a. 450-888) whose activity is inhibited upon SSD binding by cholesterol. Numerous studies have implicated the involvement of HMG-CoA reductase and mevalonate pathway in carcinogenesis.

Specificity

Clone IgG-A9 targets an epitope within HMC-CoA reductase catalytic domain (a.a. 450 887).

Immunogen

Membrane preapration from compactin-adapted CHO cells (Liscum, L., et al. (1983). J. Biol. Chem. 258(13) 8450-8455).

Application

Detect HMG CoA Reductase using this mouse monoclonal Anti-HMG-CoA Reductase, clone IgG-A9, Cat. No. MABS1233, validated for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:100 dilution from a representative lot detected in 15 µg of membrane extract from CHO7 cells treated overnight with 0.01 mM compactin (Courtesy of Linda Donnelly, Department of Molecular Genetics, UT Southwestern Medical Center, Dallas, TX, USA).

Immunocytochemistry Analysis: A representative lot detected HMG CoA reductase-positive ER membrane structures by indirect immunofluorescence staining of paraformaldehyde-fixed, 0.1% Triton X-100-permeabilized CHO-K1 cells cultured with compactin, mevalonate, and MG-132, in the presence or absence of 25-hydroxycholesterol (25-HC) (Hartman, I.Z., et al. (2010). J. Biol. Chem. 285(25):19288-19298).

Immunoprecipitation Analysis: A representative lot immunodepleted HMC-CoA reductase activity from compactin-adapted CHO cell (UT-1) extract (Liscum, L., et al. (1983). J. Biol. Chem. 258(13) 8450-8455).

Western Blotting Analysis: A representative lot detected 25-hydroxycholesterol (25-HC) treatment-induced HMC-CoA reductase dislocation from ER membrane to the cytosol and lipid droplet in CHO-K1 cells cultured with lipoprotein-deficient serum in the presence of compactin, MG-132, with or without mevalonate. Cellular VCP knockdown prevented sterol-induced HMC-CoA reductase cytosolic, but not lipid droplet, translocation (Hartman, I.Z., et al. (2010). J. Biol. Chem. 285(25):19288-19298).

Western Blotting Analysis: A representative lot detected 25-hydroxycholesterol (25-HC) treatment-induced ubiquitination of HMC-CoA reductase in SV-589 human fibroblasts cultured with compactin and mevalonate in the presence of proteasome inhibitor MG-132. Cellular knockdown of gp78A, but not gp78B or Hrd1, reduced 25-HC-induced HMC-CoA reductase ubiquitination (Song, B.L., et al. (2005). Mol. Cell. 19(6):829-840).

Western Blotting Analysis: A representative lot detected 25-hydroxycholesterol (25-HC) treatment-induced degradation of HMC-CoA reductase in SV-589 human fibroblasts cultured with compactin and mevalonate. Cellular knockdown of gp78A, VCP, or Insig-1/-2 suppressed sterol-induced HMC-CoA reductase degradation (Song, B.L., et al. (2005). Mol. Cell. 19(6):829-840).

Western Blotting Analysis: A representative lot detected HMC-CoA reductase in both membrane and nuclear fractions from HEK-293S cells cultured with compactin and mevalonate. Additional treatment with 25-hydroxycholesterol (25-HC) resulted in HMC-CoA reductase degradation (Sever, N., et al. (2003). Mol. Cell. 11(1):25-33).

Western Blotting Analysis: A representative lot detected an upregulated HMC-CoA reductase expression in compactin-adapted CHO cells (UT-1), as well as a loss of HMC-CoA reductase expression in UT-1 cells cultured in the presence of LDL (Liscum, L., et al. (1983). J. Biol. Chem. 258(13) 8450-8455).

Western Blotting Analysis: A representative lot detected a greater HMC-CoA reductase upregulation in liver microsome preparation from rats on diet supplemented with cholestyramine and mevinolin than with cholestyramine alone, while supplementation with cholesterol in addition to cholestyramine and mevinolin downregulated liver HMC-CoA reductase (Liscum, L., et al. (1983). J. Biol. Chem. 258(13) 8450-8455).

Quality

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1 .

Target description

~92 kDa observed. 97.08 kDa (hamster), 97.48/92.02/99.75 kDa (human isoform 1/2/3 (HMGCR-1b)), 96.69 kDa (rat) calculated.. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiu-Qing Li et al.
Journal of virology, 97(6), e0041223-e0041223 (2023-05-31)
Pseudorabies virus (PRV) is a double-stranded DNA virus that causes Aujeszky's disease and is responsible for economic loss worldwide. Transmembrane protein 41B (TMEM41B) is a novel endoplasmic reticulum (ER)-localized regulator of autophagosome biogenesis and lipid mobilization; however, the role of
Peng-Wei Yu et al.
Microbiology spectrum, 10(2), e0227621-e0227621 (2022-04-12)
Porcine reproductive and respiratory syndrome virus (PRRSV) infection leads to late-term reproductive failure and respiratory illness that affect the global swine industry. Epigallocatechin gallate (EGCG) is a polyphenolic compound from green tea that exerts antiviral activity against diverse viruses. This
Ankit Sharma et al.
PloS one, 14(1), e0209435-e0209435 (2019-01-10)
Metformin, a widely prescribed anti-diabetic drug, shows anticancer activity in various cancer types. Few studies documented that there was a decreased level of LDL and total cholesterol in blood serum of metformin users. Based on these views, this study aimed
Dönem Avci et al.
The Journal of biological chemistry, 300(2), 105644-105644 (2024-01-14)
Intramembrane proteolysis regulates important processes such as signaling and transcriptional and posttranslational abundance control of proteins with key functions in metabolic pathways. This includes transcriptional control of mevalonate pathway genes, thereby ensuring balanced biosynthesis of cholesterol and other isoprenoids. Our
Tsuyoshi Waku et al.
iScience, 24(10), 103180-103180 (2021-10-21)
Lipids, such as cholesterol and fatty acids, influence cell signaling, energy storage, and membrane formation. Cholesterol is biosynthesized through the mevalonate pathway, and aberrant metabolism causes metabolic diseases. The genetic association of a transcription factor NRF3 with obesity has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service