Skip to Content
Merck
All Photos(2)

Documents

MABN41

Sigma-Aldrich

Anti-Sodium channel Nav1.7 Antibody, clone N68/6

clone N68/6, from mouse

Synonym(s):

sodium channel, voltage-gated, type IX, alpha subunit, Sodium channel protein type IX subunit alpha, Voltage-gated sodium channel subunit alpha Nav1.7, Peripheral sodium channel 1, sodium channel, voltage-gated, type IX, alpha polypeptide, Nav1.7, sodium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

N68/6, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Voltage-gated sodium channel subunit alpha Nav1.7 (Nav1.7) is a multi-pass membrane protein containing a single IQ domain that belongs to the sodium channel family. NAV1.7 localizes to the terminal ends of sensory neurons and is believed to have a role in inflammatory pain development and mechanisms. It serves to regulate excitable membrane voltage-dependent Na+ ion permeability. There is a strong probability that Nav1.7 is involved in the transmission of nociceptive information, a suggested involvement supported by its role in a variety of chronic pain neuropathies induced by damage to the peripheral nerves. Nav1.7 is predominantly observed in dorsal root ganglion, and sensory and sympathetic neurons. Lower levels of the protein have been observed in MTC cell line, C-cell carcinoma, and smooth muscle cells. Defects in Nav1.7 expression are causal to several diseases such as autosomal recessive congenital indifference pain, primary erythermalgia, and paroxysmal extreme pain disorder or PEPD.

Immunogen

Recombinant protein corresponding to human Nav1.7.

Application

Research Category
Neuroscience

Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Sodium channel Nav1.7 Antibody, clone N68/6 is validated for use in IH, WB for the detection of Sodium channel Nav1.7.
Western Blot Analysis: A previous lot was used by an independent laboratory in HEK293 cell lysate. (James Trimmer, UC Davis/NIH NeuroMab Facility, Department of Neurobiology, Physiology and Behavior, UC Davis, Davis CA 95616-8519. neuromab@ucdavis.edu.)

Quality

Evaluated by Immunohistochemistry in rat cerebellum tissue.

Immunohistochemistry Analysis: 1:400 dilution of this antibody detected Nav1.7 in rat cerebellum tissue.

Target description

Observed at ~ 230 kDa

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat cerebellum tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assembly and validation of versatile transcription activator-like effector libraries.
Li, Y; Ehrhardt, K; Zhang, MQ; Bleris, L
Scientific Reports null
Raquel Talens-Visconti et al.
Journal of neurochemistry, 112(4), 1074-1087 (2009-12-09)
Neurite formation involves coordinated changes between the actin cytoskeleton and the microtubule network. Rho GTPases are clearly implicated in several aspects of neuronal development and function. Indeed, RhoA is a negative regulator of neurite outgrowth and its effector Rho-kinase mediates
Liu Yang et al.
Neuron, 93(4), 806-821 (2017-02-07)
The current knowledge about heat nociception is mainly confined to the thermosensors, including the transient receptor potential cation channel V1 expressed in the nociceptive neurons of dorsal root ganglion (DRG). However, the loss of thermosensors only partially impairs heat nociception
Dong Liu et al.
F1000Research, 5, 2764-2764 (2016-12-19)
Identification of small and large molecule pain therapeutics that target the genetically validated voltage-gated sodium channel Na V1.7 is a challenging endeavor under vigorous pursuit. The monoclonal antibody SVmab1 was recently published to bind the Na V1.7 DII voltage sensor
Fei Dong et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(50), 9589-9601 (2020-11-12)
Itch can be induced by activation of small-diameter DRG neurons, which express abundant intracellular fibroblast growth factor 13 (FGF13). Although FGF13 is revealed to be essential for heat nociception, its role in mediating itch remains to be investigated. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service