Skip to Content
Merck
All Photos(1)

Key Documents

MABE132

Sigma-Aldrich

Anti-MyoD Antibody, clone 5F11

clone 5F11, from rat

Synonym(s):

Myoblast determination protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F11, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Myod1(17927)

General description

Myoblast determination protein 1 (MyoD1) is a myogenic transcription factor required for the growth, repair, and regeneration of muscle cells. It induces differentiation of myoblast cells into multinucleated myotubules, activating genes and proteins involved in cell cycle arrest, and differentiation. MyoD1 may also control proliferation and homeostasis during embryogenesis, in the absence of Myf5.

Immunogen

GST-tagged recombinant protein corresponding to mouse MyoD.

Application

Detect MyoD using this Anti-MyoD Antibody, clone 5F11 validated for use in Western Blotting, ICC.
Immunocytochemistry Analysis: A representative lot detected MyoD in C2C12 cells.

Western Blot Analysis: A representative lot detected MyoD in C2C12 cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blot in C2C12 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected MyoD in 10 µg of C2C12 cell lysate.

Target description

~45 kDa observed. An uncharacterized band at ~38 kDa may be observed in some cell lysates. The calculated molecular weight is 34 kDa; however, this protein is known to be glycosylated.

Linkage

Replaces: MABE514

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
C2C12 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia González et al.
PloS one, 11(5), e0154919-e0154919 (2016-05-06)
Skeletal muscle regeneration and long term maintenance is directly link to the balance between self-renewal and differentiation of resident adult stem cells known as satellite cells. In turn, satellite cell fate is influenced by a functional interaction between the transcription
Akitoshi Hara et al.
Genes to cells : devoted to molecular & cellular mechanisms, 26(7), 495-512 (2021-05-08)
Mesenchymal stem cells (MSCs) are the likely precursors of multiple lines of mesenchymal cells. The existence of bona fide MSCs with self-renewal capacity and differentiation potential into all mesenchymal lineages, however, has been unclear because of the lack of MSC-specific
Claire Kim et al.
Annals of biomedical engineering, 48(2), 893-902 (2019-12-06)
Stiffness gradient hydrogels are a useful platform for studying mechanical interactions between cells and their surrounding environments. Here, we developed linear stiffness gradient hydrogels by controlling the polymerization of gelatin methacryloyl (GelMA) via differential UV penetration with a gradient photomask.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service