Skip to Content
Merck
All Photos(1)

Key Documents

MAB8672-I

Sigma-Aldrich

Anti-HSV1-ICP8 Antibody, clone 10A3

clone 10A3, from mouse

Synonym(s):

Major DNA-binding protein, HSV1-ICP8, Infected cell protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10A3, monoclonal

species reactivity

virus

species reactivity (predicted by homology)

all

technique(s)

western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Major DNA-binding protein (UniProt: P04296) is encoded by DBP (also known as Infected cell protein 8, ICP8, UL29) gene (Gene ID: 2703458) in human herpes simplex virus 1. HSV1-ICP8 is a 128 kDa, single stranded DNA binding protein of the herpesviridae family. It is an essential replication factor for both Herpes Simplex Virus Type 1 (HSV-1) and Adeno-Associated Virus (AAV) DNA replication. It binds tightly and cooperatively to ssDNA and destabilizes dsDNA helices, promotes DNA strand transfer, and renaturation of ssDNA. ICP8 is reported to regulate viral gene expression by repressing transcription from the parental viral genome and stimulating gene expression from the progeny genome. It is known to associate in replication compartments with over 50 cellular and viral proteins-many of which are involved in DNA replication and repair, recombination, and chromatin modeling. In the absence of DNA replication, HSV1-ICP8 is found in the nuclear framework-associated structures (pre-replicative sites); however, as viral DNA replication proceeds, it migrates to globular intranuclear structures (replication compartments). (Ref.: Glauser, D.L., et al. (2007). J. Virol. 81(9): 4732-4743; Tolun, G., et al. (2013). Nucleic Acids Res. 41(11): 5927 5937).

Specificity

Clone 10A3 detects ICP-8 protein in human herpes simplex virus 1.

Immunogen

ICP8 purified from U-35-VERO cells.

Application

This mouse monoclonal Anti-HSV1-ICP8, clone 10A3 , Cat. No. MAB8672-I is validated for use in Western Blotting for the detection of Herpes Simplex Virus Infected cell protein 8.
Western Blotting Analysis: A representative lot detected HSV1-ICP8 in HeLa cells (Glauser, D.L., et. al. (2007). J Virol. 81(9):4732-43).

Quality

Evaluated by Western Blotting in using recombinant protein.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected 1.25 µg of 1CB8 recombinant protein.

Target description

~128 kDa observed; 128.35 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumio Maeda et al.
Journal of virology, 91(12) (2017-03-31)
Upon herpes simplex virus 1 (HSV-1) infection, the CD98 heavy chain (CD98hc) is redistributed around the nuclear membrane (NM), where it promotes viral de-envelopment during the nuclear egress of nucleocapsids. In this study, we attempted to identify the factor(s) involved
Ken Sagou et al.
Journal of virology, 84(4), 2110-2121 (2009-12-04)
Herpesvirus nucleocapsids assemble in the nucleus and must cross the nuclear membrane for final assembly and maturation to form infectious progeny virions in the cytoplasm. It has been proposed that nucleocapsids enter the perinuclear space by budding through the inner
Zhuoming Liu et al.
Journal of virology, 89(17), 8982-8998 (2015-06-19)
To clarify the function(s) of the herpes simplex virus 1 (HSV-1) major virion structural protein UL47 (also designated VP13/14), we screened cells overexpressing UL47 for UL47-binding cellular proteins. Tandem affinity purification of transiently expressed UL47 coupled with mass spectrometry-based proteomics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service