Skip to Content
Merck
All Photos(1)

Key Documents

MAB5780

Sigma-Aldrich

Anti-NMDAR2B Antibody

ascites fluid, clone 1E4.25A4, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

1E4.25A4, monoclonal

species reactivity

mouse, rabbit, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

NMDAR2B, N-terminal. By Western blot the antibody reacts with bands at ~170 and ~48 kDa on rat brain lystate. An additional band at ~100 kDa may be seen depending on sample and antibody concentration used.

Immunogen

Recombinant protein from rat NMDAR2B.

Application

Anti-NMDAR2B Antibody detects level of NMDAR2B & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:500-1:1,000 on rat brain lysate.

Optimal working dilutions must be determined by end user.

Physical form

Liquid

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Svensson et al.
Pain, 148(3), 473-480 (2010-01-13)
Injection of nerve growth factor (NGF) into the masseter muscle is not painful but does induce a localized, quick onset ( approximately 1h) and long-lasting mechanical sensitization in healthy human subjects. We tested the hypothesis that human NGF mechanically sensitizes
Ting Sun et al.
Neurotoxicity research, 38(4), 979-991 (2020-05-06)
Anxiety leads to a global decline in quality of life and increase in social burden. However, treatments are limited, because the molecular mechanisms underlying complex emotional disorders are poorly understood. We explored the anxiolytic effects of 8-O-acetyl shanzhiside methylester (8-OaS)
Han Zhao et al.
Brain and behavior, 8(7), e01004-e01004 (2018-06-02)
It is known that an interruption of acoustic input in early life will result in abnormal development of the auditory system. Here, we further show that this negative impact actually spans beyond the auditory system to the hippocampus, a system
Zhao-hui Xu et al.
PloS one, 7(10), e48741-e48741 (2012-11-03)
Fragile X syndrome is a common inherited form of mental retardation caused by the lack of fragile X mental retardation protein (FMRP) because of Fmr1 gene silencing. Serotonin (5-HT) is significantly increased in the null mutants of Drosophila Fmr1, and
Qi-Xin Shi et al.
Molecular brain, 10(1), 38-38 (2017-08-13)
The G protein-coupled receptor 55 (GPR55) is a novel cannabinoid receptor, whose exact role in anxiety remains unknown. The present study was conducted to explore the possible mechanisms by which GPR55 regulates anxiety and to evaluate the effectiveness of O-1602

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service