Skip to Content
Merck
All Photos(1)

Key Documents

MAB4147

Sigma-Aldrich

Anti-MRP1 Antibody, clone MRPm5

clone MRPm5, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

MRPm5, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

Gene Information

human ... ABCC1(4363)

Related Categories

Specificity

This antibody reacts with an internal epitope of MRP1, a 180-195 kDa transmembrane transporter protein overexpressed in various human non-P-glycoprotein MDR tumor cell lines. The antibody does not cross-react with the human MDR1 and MDR3 gene products.

Immunogen

Bacterial fusion protein of MRP1 containing amino acids 986-1204 of the protein.

Application

Detect MRP1 using this Anti-MRP1 Antibody, clone MRPm5 validated for use in FC, WB, IH, IH(P).
Research Category
Metabolism
Research Sub Category
Toxicology & Drug Resistance
Western blotting

Immunohistochemistry: 1:20 on acetone fixed frozen tissue sections or paraffin-embedded tissue sections

Flow Cytometry

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified from tissue culture supernatant, serum-free. Liquid containing 0.7% BSA and 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. May be stored at 2-8°C for short term use. Avoid repeated freeze-thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of the multidrug resistance-associated protein (MRP) gene in primary non-small-cell lung cancer.
Nooter, K, et al.
Annals of Oncology, 7, 75-81 (1996)
Zhongxing Liang et al.
Biochemical pharmacology, 79(6), 817-824 (2009-11-04)
Multidrug resistance-associated protein (MRP-1/ABCC1) transports a wide range of therapeutic agents and may play a critical role in the development of multidrug resistance (MDR) in tumor cells. However, the regulation of MRP-1 remains controversial. To explore whether miRNAs are involved
Zhongxing Liang et al.
Pharmaceutical research, 28(12), 3091-3100 (2011-08-20)
To explore whether miR-19 is involved in the regulation of multidrug resistance (MDR), one of the main causes of breast cancer mortality, and modulates sensitivity of tumor cells to chemotherapeutic agents. We analyzed miRNA expression levels in three MDR cell
Tissue distribution of the multidrug resistance protein.
Flens, M J, et al.
The American Journal of Pathology, 148, 1237-1247 (1996)
Prognostic significance of the immunohistochemical expression of O(6)-methylguanine-DNA methyltransferase, P-glycoprotein, and multidrug resistance protein-1 in glioblastomas.
Takao Nakagawa, Kazunori Ido, Takahiro Sakuma, Hiroaki Takeuchi, Kazufumi Sato et al.
Neuropathology : Official Journal of the Japanese Society of Neuropathology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service