Skip to Content
Merck
All Photos(3)

Documents

MAB3574-C

Sigma-Aldrich

Anti-Vinculin Antibody, clone VIIF9 (7F9), Ascites Free

clone VIIF9 (7F9), from mouse

Synonym(s):

Metavinculin, Meta-vinculin, MV, Vinculin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

VIIF9 (7F9), monoclonal

species reactivity

rat, mouse, rabbit, human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)

Related Categories

General description

Metavinculin (MV) and its a.a. 916-983 truncated splice variant Vinculin (UniProt P18206) are cytoskeleton proteins encoded by the VCL (also known as MVCL, CMD1W, CMH15) gene (Gene ID 7414) in human. The N-terminal globular head domain (a.a. 2-835) of Vinculin/VM contains binding sites for talin and α-actinin as well as a tyrosine phosphorylation site, while their C-terminal Tail region (a.a. 879-1134 of VM; a.a. 879-1066 of Vinculin) contains binding sites for F-actin, paxillin, and lipids. Vinculin is associated with focal adhesion and adherens junctions, which are complexes that nucleate actin filaments and crosslinkers between the external medium, plasma membrane, and actin cytoskeleton. The loss of vinculin disrupts focal adhesion complex formation and prevents cell adhesion and spreading, accompanied by reduced stress fiber formation and inhibition of lamellipodia extension. Smooth muscles and skeletal muscles in their well-differentiated, contractile state co-express vinculin and meta-vinculin. Both vinculin isoforms co-localize in muscular adhesive structures, such as dense plaques in smooth muscles, intercalated discs in cardiomyocytes, and costameres in skeletal muscles.

Specificity

Clone VII F9 (7F9) reacted equally with human vinculin and meta-vinculin by immunoblotting and recognized all vinculin (α, α′, β, γ) and meta-vinculin (α & β) variants resolved by isoelectrofocusing (Glukhova, M.A., et al. (1990). J. Biol. Chem. 265:13042-13046).

Immunogen

Human uterus meta-vinculin.

Application

Anti-Vinculin Antibody, clone VIIF9 (7F9), Ascites Free
is an antibody against Vinculin for use in Western Blotting
, Immunohistochemistry (Paraffin, Immunofluorescence

Immunocytochemistry.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Viculin in 10 µg of A431 cell lysate.
Immunohistochemistry Analysis: An 1:1,000 dilution from a representative lot detected Viculin in human kidney and colon tissue sections.
Western Blotting Analysis: A representative lot detected vinculin in HEK293 and HepG2 lysates (Veronese, A., et al. (2011). Proc. Natl. Acad. Sci. U. S. A. 108(12):4840-4845).
Western Blotting Analysis: Clone VII F7 reacted equally with human vinculin and meta-vinculin by immunoblotting and recognized all human vinculin (α, α′, β, γ) and meta-vinculin (α & β) variants resolved by isoelectrofocusing (Glukhova, M.A., et al. (1990). J. Biol. Chem. 265:13042-13046).
Immunofluorescence Analysis: A representative lot immunostained the focal adhesion protein vinculin in acetone-fixed frozen mouse skeletal and cardiac muscle sections by fluorescent immunohistochemistry (Belkin, A.M., et al. (1996). J. Cell Biol. 132(1-2):211-226).
Immunocytochemistry Analysis: A representative lot detected an increased average size and number of focal adhesions per cell following miR-200-mediated genes knockdown by fluorescent immunocytochemistry staining of paraformaldehyde-fixed, Triton X-100-permeabilized MDA-MB-231 human breast cancer cells (Bracken, C.P., et al. (2014). EMBO J. 33(18):2040-2056).
Immunocytochemistry Analysis: A representative lot immunostained the focal adhesion protein vinculin in paraformaldehyde-fixed, Triton X-100-permeabilized human fibroblasts by fluorescent immunocytochemistry (Addad, S., et al. (2011). Mar. Drugs. 9(6):967-983).
Immunocytochemistry Analysis: A representative lot immunostained the focal adhesion protein vinculin in fixed human intestinal epithelial cells (HIECs) by fluorescent immunocytochemistry (Gagné , D., et al. (2010). J. Cell Physiol. 222(2):387-400).
Immunocytochemistry Analysis: Representative lots immunostained the focal adhesion protein vinculin in paraformaldehyde-fixed, Triton X-100-permeabilized REF52 rat embryonic fibroblasts, mouse skin fibroblasts, rabbit chondrocytes, and human MCF-7 cells by fluorescent immunocytochemistry (Akimov, S.S., et al. (2000). J. Cell Biol. 148(4):825-838; Jurjus, R.A., et al. (2008). Wound Repair Regen. 16(5):649-660; Yamamoto, K., et al. (2007). Biomaterials. 28(10):1838-1846; Poppe, M., et al. (2007). Oncogene. 26(24):3462-3472).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Viculin in 10 µg of HepG2 cell lysate.

Target description

~130 kDa observed. Target band size appears larger than the calculated molecular weights of 116.7 kDa (Vinculin; isoform 1) and 123.80 kDa (Metainculin; isoform 2).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Wen Chai et al.
PloS one, 11(9), e0162853-e0162853 (2016-09-16)
The development and utilization of three-dimensional cell culture platforms has been gaining more traction. Three-dimensional culture platforms are capable of mimicking in vivo microenvironments, which provide greater physiological relevance in comparison to conventional two-dimensional cultures. The majority of three-dimensional culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service