Skip to Content
Merck
All Photos(1)

Key Documents

MAB3226

Sigma-Aldrich

Anti-Cytokeratin 7 Antibody, clone RCK105

clone RCK105, Chemicon®, from mouse

Synonym(s):

Anti-CD20, Anti-CK-20, Anti-CK20, Anti-K20, Anti-KRT21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RCK105, monoclonal

species reactivity

hamster, canine, human, feline, pig, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

dog ... Krt7(477602)
human ... KRT7(3855)
mouse ... Krt7(110310)
rat ... Krt7(300242)

General description

Keratin, type II cytoskeletal 7 (UniProt: P08729; also known as Cytokeratin-7, CK-7, Keratin-7, K7, Sarcolectin, Type-II keratin Kb7) is encoded by the KRT7 (also known as SCL) gene (Gene ID: 3855) in human. Cytokeratins are the intermediate filament proteins of epithelia that display high degree of molecular diversity. They are heteropolymeric filaments formed by pairing of type I and type II keratins that are expressed in a highly specific patterns depending on the epithelial type and stage of cellular differentiation. They contain a central rod domain of about 310 amino acids with α-helical conformation bordered by non-helical head and tail domains of variable length. The head domain consists of subdomains V1 and H1. The central a-helical rod domain is composed of subdomains 1A, 1B, 2A, and 2B connected by the linkers L1, L12, and L2. The tail domain then consists of subdomains H2 and V2. They provide stability between epithelial cells and their attachment to basement membrane. Keratins also serve as tumor markers and epithelial differentiation markers. Multiple human epithelial keratins have been resolved with 2D-gels electrophoresis. These can be divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. The acidic keratins have molecular weights that range from 40 to 55 kDa. The basic keratins have molecular weights that range from 56 to 70 kDa. Members of the acidic and basic subfamilies are reported to be present in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and the disease state. Keratin 7 is expressed in cultured epidermal, bronchial, and mesothelial cells. It is observed throughout the glandular cells in the junction between stomach and esophagus but is absent in esophagus. Its expression is also observed in a variety of carcinomas and in tumor cells of mammary and extramammary Paget disease. Keratin-7 expression can be used to distinguish ovarian and gastrointestinal carcinomas. (Ref.: Chu, P., et al. (2000). Modern Pathol. 13(9); 962-972; Kaba, A., et al. (1999). Biochimie. 81(7); 709-715).

Specificity

MAB3226 reacts with a subgroup of glandular epithelial and their tumors, as well as transitional epithelium and transitional carcinoma. It reacts with a 58 kDa protein corresponding to cytokeratin 7.

Immunogen

Cytokeratins from the human bladder carcinoma cell line T24.

Application

Anti-Cytokeratin 7 Antibody, clone RCK105 is an antibody against Cytokeratin 7 for use in FC, WB, IC, IH.
Research Category
Cell Structure
Research Sub Category
Cytokeratins
Western blot: 57-58kDa band in reduce westerns

Immunohistochemistry on frozen tissue sections.

Immunocytochemistry

Flow cytometry

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: MABT150

Physical form

Format: Purified
Liquid in buffer with 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ichiro Onoyama et al.
The Journal of clinical investigation, 121(1), 342-354 (2010-12-03)
E3 ubiquitin ligase complexes of the SCF type consist of ring-box 1 (Rbx1), cullin 1 (Cul1), S-phase kinase-associated protein 1 (Skp1), and a member of the F-box family of proteins. The identity of the F-box protein determines the substrate specificity
David Campard et al.
Gastroenterology, 134(3), 833-848 (2008-02-05)
Umbilical cord matrix stem cells (UCMSCs) are able to differentiate into mesodermal and ectodermal lineages. The present study investigates the differentiation potential of human UCMSCs into hepatic lineage. We isolated human UCMSCs and characterized them in vitro by measuring their
Hsiao-Fan Lo et al.
The Journal of pathology, 241(3), 324-336 (2016-12-06)
The transcription factor glial cells missing 1 (GCM1) regulates trophoblast differentiation and function during placentation. Decreased GCM1 expression is associated with pre-eclampsia, suggesting that abnormal expression of GCM1 target genes may contribute to the pathogenesis of pregnancy complications. Here we
Andreja Erman et al.
International journal of molecular sciences, 22(12) (2021-07-03)
Non-muscle-invasive bladder cancer is the most common form of bladder cancer. The main problem in managing bladder tumors is the high recurrence after the transurethral resection of bladder tumors (TURBT). Our study aimed to examine the fate of intravesically applied
Miki Nishio et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(1), E71-E80 (2015-12-25)
Mps One Binder Kinase Activator (MOB)1A/1B are core components of the Hippo pathway that coactivate large tumor suppressor homolog (LATS) kinases. Mob1a/1b double deficiency in mouse liver (LMob1DKO) results in hyperplasia of oval cells and immature cholangiocytes accompanied by inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service