Skip to Content
Merck
All Photos(1)

Documents

MAB3120

Sigma-Aldrich

Anti-Cre Recombinase Antibody, clone 2D8

ascites fluid, clone 2D8, Chemicon®

Synonym(s):

Anti-Cre Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2D8, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Escherichia coli ... Cre(2777477)

General description

Cre-recombinase is a 38 kDa DNA recombinase derived from the P1 bacteriophage. It is highly specific for a 34 bp DNA sequence (loxP) found in P1 DNA. It catalyzes site-specific recombination between two 34-base-pair LOXP sites. Its role is to maintain the phage genome as a monomeric unit-copy plasmid in the lysogenic state. It is homotetramer when bound to DNA and belongs to the "phage" integrase family. Anti-Cre Recombinase Antibody can be used to determine the success of gene manipulations in transgenic animals.

Specificity

Cre recombinase. The epitope is located between aa 77 and 343.

Immunogen

Cre recombinase (aa 77 to 343) fusion protein

Application

Anti-Cre Recombinase Antibody, clone 2D8 is a well characterized monoclonal antibody. Proven to reliably detect Cre Recombinase, this mAb is validated for use in ELISA, IC, IF, IH & WB and is backed by multiple publications.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blot (natural and recombinant protein):
A 1:500-1:5,000 dilution was used in a previous lot. Natural Cre recombinase from Bacteriophage P1 migrates at 38.5 kDa by SDS-PAGE.

Immunofluorescence:
A previous lot of this antibody was used on IF.

Immunohistochemistry:
A 1:500-1:5,000 dilution of a previous lot was used in IH.

ELISA:
A 1:500-1:5,000 dilution of a previous lot was used in ELISA.

Optimal working dilutions must be determined by the end user.

Target description

38 kDa

Physical form

Ascites fluid. Contains no preservative
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
HeLa Nuclear Extract

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Xiang Liu et al.
Developmental biology, 368(2), 294-303 (2012-06-05)
The epithelium of mammalian tongue hosts most of the taste buds that transduce gustatory stimuli into neural signals. In the field of taste biology, taste bud cells have been described as arising from "local epithelium", in distinction from many other
Extrasynaptic GABAA receptors in mediodorsal thalamic nucleus modulate fear extinction learning.
Paydar, A; Lee, B; Gangadharan, G; Lee, S; Hwang, EM; Shin, HS
Molecular Brain null
Axon position within the corpus callosum determines contralateral cortical projection.
Zhou, J; Wen, Y; She, L; Sui, YN; Liu, L; Richards, LJ; Poo, MM
Proceedings of the National Academy of Sciences of the USA null
Takashi Umehara et al.
Endocrinology, 157(12), 4899-4913 (2016-10-13)
Adult Leydig cells are derived from proliferating stem/progenitor Leydig cells in the infant testis and subsequent differentiation to steroidogenic cells in adult mice. Leydig cell proliferation in the infant testis occurs primarily in response to increased levels of LH that
Arlene A Hirano et al.
eNeuro, 3(2) (2016-03-30)
The cellular mechanisms underlying feedback signaling from horizontal cells to photoreceptors, which are important for the formation of receptive field surrounds of early visual neurons, remain unsettled. Mammalian horizontal cells express a complement of synaptic proteins that are necessary and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service