Skip to Content
Merck
All Photos(1)

Key Documents

MAB2251

Sigma-Aldrich

Anti-Integrin β1 Antibody, clone B3B11

clone B3B11, Chemicon®, from mouse

Synonym(s):

CD29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B3B11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Specificity

Reacts with the human integrin beta1 subunit. Specificity verified by preclearing of beta1in immunoprecipitation, flow cytometry on transfectant cells displaying human beta1 integrin, and reactivity with purified beta1. Epitope lies within the same cluster recognized by MAB2250, and within amino acids 648-670 of the mature beta1 integrin chain.1

Immunogen

source is Jurkat T-leukemic cell line

Application

Anti-Integrin β1 Antibody, clone B3B11 is an antibody against Integrin β1 for use in FC, IP, WB, FUNC, IH.
Immunoprecipitation

Immunohistochemistry on frozen sections

Flow cytometry

Immunoblotting (Western)

Stimulates adhesion of cells to extracellular matrix proteins1

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Purified immunoglobulin from Protein A Sepharose chromatography. Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Sepharose is a trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumitra Mukhopadhyay et al.
Bioconjugate chemistry, 19(1), 39-49 (2007-09-12)
The integrins alpha vbeta3 and alpha vbeta5 and the membrane-spanning surface protein aminopeptidase N (APN) are highly expressed in tumor-induced angiogenesis, making them attractive targets for therapeutic intervention. Both integrins and APN recognize a broad range of peptides containing RGD
Faye Hui Chen et al.
The Journal of biological chemistry, 280(38), 32655-32661 (2005-07-30)
Cartilage oligomeric matrix protein/thrombospondin 5 (COMP/TSP5) is a major component of the extracellular matrix of the musculoskeletal system. Although COMP/TSP5 abnormalities are associated with several pathological conditions, its normal function remains unclear. This study was undertaken to delineate the function(s)
Alison McDowall et al.
The Journal of clinical investigation, 111(1), 51-60 (2003-01-04)
The adhesion receptors known as integrins perform key functions for hematopoietic cells. The platelet integrin alphaIIbbeta3 is critical in hemostasis, and the beta1 and beta2 integrins on leukocytes have many roles in cell-mediated immunity. Mutations in the beta2 subunit lead
Salvatore Condello et al.
Cancer research, 78(11), 2990-3001 (2018-03-08)
Cancer progression and recurrence are linked to a rare population of cancer stem cells (CSC). Here, we hypothesized that interactions with the extracellular matrix drive CSC proliferation and tumor-initiating capacity and investigated the functions of scaffold protein tissue transglutaminase (TG2)
Ewa Pocheć et al.
Analytical cellular pathology (Amsterdam), 2015, 324980-324980 (2015-09-05)
N-glycosylation plays an important role in the majority of physiological and pathological processes occurring in the immune system. Alteration of the type and abundance of glycans is an element of lymphocyte differentiation; it is also common in the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service