Skip to Content
Merck
All Photos(1)

Key Documents

MAB2021Z

Sigma-Aldrich

Anti-Integrin αV (ITGAV/CD51) Antibody

CHEMICON®, mouse monoclonal, AV1

Synonym(s):

CD51, VitronectinReceptor, MAB2021

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin αV Antibody, extracellular, clone AV1, azide free, culture supernatant, clone AV1, Chemicon®

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AV1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Specificity

Specific for human integrin alphaV (other species not tested).

Immunogen

Epitope: extracellular, azide free
Human integrin alphaVbeta3 heterodimer isolated from placenta.

Application

Anti-Integrin αV Antibody, extracellular, clone AV1, azide free detects level of Integrin αV & has been published & validated for use in FC & WB.
Flow cytometry: 1:5-1:50

Western Blot. Under non-reducing conditions recognizes a band at approximately 165 kDa corresponding to integrin alphaV. Does not recognize the reduced protein.

Function-blocking of integrin alphaV-mediated adhesion to ECM proteins. (1:5-1:20, in HEPES buffer pH 7.4 containing 1mM Mn++ and 1mM Mg++).

Optimal working dilutions and protocols must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Unpurified tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human metapneumovirus (HMPV) binding and infection are mediated by interactions between the HMPV fusion protein and heparan sulfate.
Chang, A; Masante, C; Buchholz, UJ; Dutch, RE
Journal of virology null
Osteopontin enhances the expression and activity of MMP-2 via the SDF-1/CXCR4 axis in hepatocellular carcinoma cell lines.
Zhang, R; Pan, X; Huang, Z; Weber, GF; Zhang, G
Testing null
Jakob Dalgaard et al.
Journal of leukocyte biology, 77(5), 689-698 (2005-02-25)
CD11c+ dendritic cells (DC) and plasmacytoid DC (PDC) are the two major DC subsets in human peripheral blood. For the purpose of immunotherapy with DC, it is important to investigate the phagocytosis of killed tumor cells by different DC subsets.
Bing-Ling Yun et al.
The Journal of biological chemistry, 291(28), 14815-14825 (2016-05-27)
Avian metapneumovirus (aMPV) fusion (F) protein mediates virus-cell membrane fusion to initiate viral infection, which requires F protein binding to its receptor(s) on the host cell surface. However, the receptor(s) for aMPV F protein is still not identified. All known
Brett G Hollier et al.
Endocrinology, 149(3), 1075-1090 (2007-12-15)
IGF-I can bind to the extracellular matrix protein vitronectin (VN) through the involvement of IGF-binding proteins-2, -3, -4, and -5. Because IGF-I and VN have established roles in tumor cell dissemination, we were keen to investigate the functional consequences of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service