Skip to Content
Merck
All Photos(1)

Key Documents

MAB1950Z

Sigma-Aldrich

Anti-Integrin α2 Antibody, clone P1E6, azide free

clone P1E6, Chemicon®, from mouse

Synonym(s):

CD49b, MAB1950

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1E6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA2(3673)

Specificity

Recognizes Alpha 2 integrin.

Application

Suitable for use in attachment inhibition assays using fibroblasts, epithelial cells, endothelial cells, and non-activated platelets on collagen types I, III, IV, VI and laminin.

Suitable for immunofluorescence using fresh frozen or acetone fixed tissues. Not recommended for traditional formalin fixed paraffin embedded tissues.

Final working dilutions must be determined by end user.
This Anti-Integrin α2 Antibody, clone P1E6, azide free is validated for use in IC, IH, FUNC for the detection of Integrin α2.

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 0.01 M phosphate buffered saline, pH 7.1, with no preservatives.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam Becker et al.
Journal of virology, 97(10), e0077023-e0077023 (2023-10-12)
Enteric adenoviruses have historically been difficult to grow in cell culture, which has resulted in lack of knowledge of host factors and pathways required for infection of these medically relevant viruses. Previous studies in non-intestinal cell lines showed slow infection
Richard A Que et al.
Journal of biomedical materials research. Part A, 106(5), 1363-1372 (2018-01-18)
Adhesion to the microenvironment profoundly affects stem cell functions, including proliferation and differentiation, and understanding the interaction of stem cells with the microenvironment is important for controlling their behavior. In this study, we investigated the effects of the integrin binding
Angara Sureshbabu et al.
Journal of cell science, 125(Pt 7), 1693-1705 (2012-02-14)
Maintenance of tissue boundaries is crucial for control of metastasis. We describe a new signalling pathway in which epithelial cell disruption can be minimised and thereby restricts epithelial-mesenchymal transgressions. This involves the release of insulin-like growth factor (IGF)-binding protein 5
Joie A Bernard-Trifilo et al.
Journal of neurochemistry, 93(4), 834-849 (2005-04-29)
Integrin class adhesion proteins are concentrated at adult brain synapses. Whether synaptic integrins engage kinase signaling cascades has not been determined, but is a question of importance to ideas about integrin involvement in functional synaptic plasticity. Accordingly, synaptoneurosomes from adult
Amy Meyer et al.
Neoplasia (New York, N.Y.), 6(4), 332-342 (2004-07-17)
Neuroblastoma (NBL) is the most common malignant disease of infancy, and children with bone metastasis have a mortality rate greater than 90%. Two major classes of proteins, integrins and growth factors, regulate the metastatic process. We have previously shown that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service