IPAKGARD1
IPAK Gard® 3/5 Pretreatment Cartridge
For Milli-Q® IQ/IX 7003/05 and Milli-Q® EQ 7008/16 water systems to protect RO membranes. This product replaces IPAKGARA1.
Synonym(s):
IPAK Gard 03/05 pretreatment pack
About This Item
Recommended Products
Quality Level
packaging
units of 1
manufacturer/tradename
Milli-Q®
storage condition
room temperature
cartridge H
39.5 cm (15.5 in.)
input
feed water nature tap water (within specifications of the water purification system)
compatibility
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
shipped in
ambient
General description
Purification technologies include activated carbon for the removal of free chlorine and a pleated filter for the removal of particulates and colloids. This pretreatment cartridge improves your system′s performance and extends its lifetime.
Application
- For Milli-Q® IQ/IX 7003/05 and EQ 7008/16 systems
- Optimally protects RO membranes from free chlorine, colloids, particles and minerals found in potable feed water.
- Improves system performance and lifetime.
Features and Benefits
- Pleated filter removes particulates and colloids, and activated carbon block removes free chlorine from potable water to ensure RO membrane performance over years.
- e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
- Supplied with a Drain Cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal.
- 63% of the parts suppliers are ISO 14001 environmental management system certified.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service