Skip to Content
Merck
All Photos(2)

Key Documents

HVHP02500

Millipore

PVDF Membrane Filter, 0.45 μm Pore Size

greener alternative

Durapore®, filter diam. 25 mm, hydrophobic

Synonym(s):

Durapore® Membrane Filter, 0.45 µm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

PVDF membrane
plain filter
white filter

Quality Level

sterility

non-sterile

feature

hydrophobic

manufacturer/tradename

Durapore®
Millipore

sustainability

Greener Alternative Product

parameter

>2.6 mL/min-cm2 water flow rate
20 L/min-cm2 air flow rate
85 °C max. temp.

diam.

25 mm

filter diam.

25 mm

thickness

125 μm

gravimetric extractables

0.5%

refractive index

n/D 1.42

matrix

Durapore®

pore size

0.45 μm pore size
75 % porosity

bubble point

≥0.6 bar, air with methanol at 23 °C

greener alternative category

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Air clarification
  • Gas and solvent filtration

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Honghe Liu et al.
Journal of the American Chemical Society, 143(19), 7368-7379 (2021-04-16)
The expansion of a hexanucleotide repeat GGGGCC (G4C2) in the C9orf72 gene is the most common cause of amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD). The G4C2 expansion leads to repeat-associated non-AUG (RAN) translation and the production of toxic
Zhongshu Zhou et al.
STAR protocols, 2(2), 100548-100548 (2021-06-08)
PAX6 is a key determinant of human neuroectoderm cell fate. Here, we describe a protocol for genome-scale CRISPR screening for use in genetically engineered human pluripotent stem cells (hPSCs). Using the germ layer reporter PAX6 and an inducible CRISPR/Cas9 knockout
Qing Tang et al.
The EMBO journal, 40(19), e107204-e107204 (2021-08-27)
Lysosomes are key organelles maintaining cellular homeostasis in health and disease. Here, we report the identification of N-deacetylase and N-sulfotransferase 3 (NDST3) as a potent regulator of lysosomal functions through an unbiased genetic screen. NDST3 constitutes a new member of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service