Skip to Content
Merck
All Photos(2)

Documents

HSTCMAG-28SK

Millipore

MILLIPLEX® Human High Sensitivity T Cell Panel - Immunology Multiplex Assay

Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Synonym(s):

Human high-sensitivity cytokine multiplex kit, Luminex® human high-sensitivity cytokine immunoassay, Millipore human high-sensitivity cytokine panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 98-107%
sensitivity: 0.11-8.17 pg/mL
(Overnight Incubation minDC)

standard curve range: 0.18-750 pg/mL
(IL-6)

standard curve range: 0.24-1,000 pg/mL
(IL-13)

standard curve range: 0.24-1,000 pg/mL
(IL-21)

standard curve range: 0.31-1,250 pg/mL
(IL-8)

standard curve range: 0.31-1,250 pg/mL
(MIP-1α)

standard curve range: 0.37-1,500 pg/mL
(IL-7)

standard curve range: 0.45-1,750 pg/mL
(TNFα)

standard curve range: 0.49-2,000 pg/mL
(IL-1β)

standard curve range: 0.49-2,000 pg/mL
(IL-12 (p70))

standard curve range: 0.49-2,000 pg/mL
(IL-2)

standard curve range: 0.49-2,000 pg/mL
(IL-5)

standard curve range: 0.61-2,500 pg/mL
(IFNγ)

standard curve range: 0.61-2,500 pg/mL
(MIP-3α)

standard curve range: 0.73-3,000 pg/mL
(IL-17A)

standard curve range: 0.92-3,750 pg/mL
(MIP-1β)

standard curve range: 1.22-5,000 pg/mL
(GM-CSF)

standard curve range: 1.46-6,000 pg/mL
(IL-10)

standard curve range: 1.46-6,000 pg/mL
(ITAC)

standard curve range: 1.83-7,500 pg/mL
(IL-4)

standard curve range: 18.3-75,000 pg/mL
(Fractalkine)

standard curve range: 7.93-32,500 pg/mL
(IL-23)

inter-assay cv: <15%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Cytokines are immunomodulatory polypeptides that play key roles in both adaptive and innate immune responses. A generic term, “cytokines” includes myokines (produced by muscle cells), lymphokines (produced by activated Th cells), interleukins (acting as mediators between T cells) and chemokines (responsible for T-cell migration). One of the regulatory mechanisms of the immune system, cytokines act at the recognition, activation, or effector phases of an immune response, modulating the development and functional activities of the subtypes of T cells, B cells and myeloid cells. Consequently, research involving cytokines plays a significant role in achieving a deeper understanding of the immune system and its multi-faceted response to most antigens, especially those responses that make up the inflammatory process. Low levels of inflammation are involved in many clinical and sub-clinical disease states, such as autoimmune disease, cardiovascular disease, diabetes, neurological disorders, and cancer. Measuring picogram levels of cytokines, therefore, is critical for understanding the pathogenesis of these diseases.

The MILLIPLEX® Human High Sensitivity T Cell Magnetic Bead Panel is to be used for the simultaneous quantification of any or all of the following analytes in human plasma, serum, and cell/tissue culture supernatant samples: Fractalkine, GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-13, IL-17A, IL-21, IL-23, ITAC, MIP-1α, MIP-1β, MIP-3α
and TNFα.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There is no detectable cross-reactivity within the panel.
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: Fractalkine, GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-13, IL-17A, IL-21, IL-23, I-TAC, MIP-1α, MIP-1β, MIP-3α, TNF-α
  • Because there are unique features concerning sample prep and assay set-up, please read the entire protocol before running the assay.
  • Recommended Sample type: Serum, plasma or tissue/cell lysate and culture supernatant
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology
  • Research Subcategory: Immune Response, Metabolic Disorders, Chronic Inflammatory Disease, Cardiovascular Disease, Neuroscience

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Linkage

Replaces: HSTCMAG-28SK; HSCYTMAG-60SK

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: Please see kit protocol for individual assay sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matteo Ravaioli et al.
Scientific reports, 10(1), 6063-6063 (2020-04-10)
With the aim to explore innovative tools for organ preservation, especially in marginal organs, we hereby describe a clinical trial of ex-vivo hypothermic oxygenated perfusion (HOPE) in the field of liver (LT) and kidney transplantation (KT) from Extended Criteria Donors
Anne H Krog et al.
Vascular health and risk management, 12, 371-378 (2016-10-08)
Minimally invasive surgical techniques have been shown to reduce the inflammatory response related to a surgical procedure. The main objective of our study was to measure the inflammatory response in patients undergoing a totally laparoscopic versus open aortobifemoral bypass surgery.
Malcolm Sanderson et al.
Sports (Basel, Switzerland), 8(12) (2020-12-05)
Swimming is a popular youth sport that is considered beneficial for cardiovascular fitness. However, the potential inflammatory outcomes of high intensity swimming in younger swimmers are unclear, as is the response of irisin, a myokine released during exercise with anti-inflammatory
Bruna M Ascoli et al.
International journal of bipolar disorders, 7(1), 13-13 (2019-06-04)
Innate immune system dysfunction has been recognized as an important element in the pathophysiology of bipolar disorder (BD). We aimed to investigate whether there are differences in the response of macrophages derived from patients in the early stages and late
B Karshikoff et al.
Brain, behavior, and immunity, 57, 38-46 (2016-04-09)
Low-grade systemic inflammation has been implicated in chronic pain, as well as in comorbid diseases like depression and fatigue. We have previously shown that women's pain perception and regulation is more affected by systemic inflammation than that of men. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service