GF021
Stem Cell Factor Protein, Recombinant human
The Stem Cell Factor (SCF) Protein, Recombinant human, is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis.
Synonym(s):
SCF, Mast Cell Growth Factor, MGF, c-kit Ligand
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77
Recommended Products
biological source
human
Quality Level
recombinant
expressed in E. coli
Assay
>98% (SDS-PAGE and HPLC)
specific activity
≥5 x 10e5 U/mg
manufacturer/tradename
Chemicon®
impurities
<0.1 ng/μg Endotoxin (of SCF; 1EU/μg)
input
sample type hematopoietic stem cell(s)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... KITLG(4254)
General description
Product Source: E.coli
Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures and has been shown to act synergistically with colony stimulating factors. Recombinant human SCF is an 18.4 kDa protein containing 165 amino acid residues corresponding to the soluble secreted form of SCF.
Application
For most in vitro applications, SCF exerts its biological activity in the concentration range of 0.2 to 20.0 ng/mL. Responding cells are (partial list): Stem cells, BM precursor cells.
Physical form
The sterile filtered solution was lyophilized from 10mM Acetic acid.
Storage and Stability
The lyophilized powder, though stable for a few weeks at room temperature, is best stored at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can be further diluted into other aqueous buffers and stored at 2-8°C for 1 week or -20°C for future use. Reconstituted SCF should be stored in undiluted aliquots at -20°C. Avoid repeated freeze/thaw cycles.
Analysis Note
Specific Activity: Chemicon′s human SCF is fully biologically active when compared to standards.The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 2 ng/mL
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Qisheng Zuo et al.
EMBO reports, 23(1), e52491-e52491 (2021-11-09)
The unique characteristics of chicken primordial germ cells (PGCs) provide potential strategies for transgenic animal generation; however, insufficient PGC availability has limited their application. Regulation of bone morphogenic protein 4 (BMP4), a crucial factor for PGCs formation, may provide new
Qisheng Zuo et al.
Journal of cell science, 134(3) (2021-01-15)
The unique developmental characteristics of chicken primordial germ cells (PGCs) enable them to be used in recovery of endangered bird species, gene editing and the generation of transgenic birds, but the limited number of PGCs greatly limits their application. Studies
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service