Skip to Content
Merck
All Photos(1)

Key Documents

FX0440

Supelco

Formic acid

98%, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Formic acid, Methanoic Acid,Aminic Acid, Aminic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-579-1
Pricing and availability is not currently available.

Product Name

Formic Acid, 98%, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

grade

ACS reagent

Quality Level

Agency

meets USP/NF

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

1004 °F

expl. lim.

57 %

impurities

≤0.05% Acetic acid (CH3COOH)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 10 g/L in H2O)

pKa (20 °C)

3.7

bp

100-101 °C (lit.)
101 °C/1013 hPa

mp

8.2-8.4 °C (lit.)

transition temp

flash point 49.5 °C

density

1.22 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
sulfate (SO42-): ≤5 ppm
sulfite (SO32-): ≤0.001%

cation traces

Ag: ≤0.02 ppm
Al: ≤0.05 ppm
Ba: ≤0.05 ppm
Be: ≤0.02 ppm
Bi: ≤0.1 ppm
Ca: ≤0.2 ppm
Cd: ≤0.05 ppm
Co: ≤0.02 ppm
Cr: ≤0.05 ppm
Cu: ≤0.02 ppm
Fe: ≤2 ppm
Ge: ≤0.05 ppm
K: ≤0.1 ppm
Li: ≤0.02 ppm
Mg: ≤0.5 ppm
Mn: ≤0.05 ppm
Mo: ≤0.02 ppm
NH4+: ≤0.001%
Na: ≤0.5 ppm
Ni: ≤0.05 ppm
Pb: ≤0.02 ppm
Sr: ≤0.02 ppm
Ti: ≤0.1 ppm
Tl: ≤0.05 ppm
V: ≤0.05 ppm
Zn: ≤0.05 ppm
Zr: ≤0.1 ppm
heavy metals (as Pb): ≤0.001%

SMILES string

OC=O

shipped in

ambient

storage temp.

room temp

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Acetic acid (CH3COOH): 0.05% max
Aluminum (Al): 0.05ppm max
Ammonium (NH4): 0.001% max
Assay: 98% min
Barium (Ba): 0.05ppm max
Beryllium (Be): 0.02ppm max
Bismuth (Bi): 0.1ppm max
Cadmium (Cd): 0.05ppm max
Calcium (Ca): 0.2ppm max
Chloride (Cl): 5ppm max
Chromium (Cr): 0.05ppm max
Cobalt (Co): 0.02ppm max
Color (APHA): 10 max
Copper (Cu): 0.02ppm max
Dilution test: To pass test
Evaporation residue: 0.001% max
Germanium (Ge): 0.05ppm max
Heavy metals (as Pb): 0.001% max
Iron (Fe): 2ppm max
Lead (Pb): 0.02ppm max
Lithium (Li): 0.02ppm max
Magnesium (Mg): 0.5ppm max
Manganese (Mn): 0.05ppm max
Molybdenum (Mo): 0.02ppm max
Nickel (Ni): 0.05ppm max
Potassium (K): 0.1ppm max
Silver (Ag): 0.02ppm max
Sodium (Na): 0.5ppm max
Strontium (Sr): 0.02ppm max
Sulfate (SO4): 5ppm max
Sulfite (SO3): 0.001% max
Thallium (Tl): 0.05ppm max
Titanium (Ti): 0.1ppm max
Vanadium (V): 0.05ppm max
Zinc (Zn): 0.05ppm max
Zirconium (Zr): 0.1ppm max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service