Skip to Content
Merck
All Photos(2)

Documents

EZFITBASE3

Millipore

EZ-Fit® Manifold

3-place manifold base for filtration application, stainless steel

Synonym(s):

EZ-Fit® Manifold base, EZ-Fit® manifold base for EZ-Stream® vacuum pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

316L stainless steel

Sterilization Compatibility

autoclavable compatible

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

EZ-FIT® vacuum manifold is a state-of-the-art system that supports sample filtration. Millipore glass, plastic or stainless-steel filter holders can be fitted with standard No. 8 perforated stoppers over holder outlets. Different heads fit the disposable consumables, as well as the reusable funnels. The manifolds optimize the workflow and control the contamination risks, and thereby avoid false-positive results during contamination tests. The design of the 2-way valve prevents accidental touching of the filtration head while opening or closing the valve. The liquid back-flow from the vacuum system is avoided by the check valve. The low height design offers better ergonomics for use in laminar flow hoods.

Application

EZ-FIT® vacuum manifolds are used as vacuum pump accessories to filter the test samples for general microbiological analysis, bioburden testing (pharmaceuticals, food, and beverage) and water monitoring. It supports the filtration of three test samples.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service