Skip to Content
Merck
All Photos(3)

Key Documents

ESA0010L2CLP5

Millipore

Mobius® Essential 2D Assembly Bag

10L PureFlex Film, 2D Bag

Synonym(s):

Mobius® Essential Assembly, 10 L, Bag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103801

material

PureFlex bag

Quality Level

Agency

complies with ISO 9001

product line

Mobius® Essential

technique(s)

buffer exchange: suitable
media fill: suitable

H

25 m

W

12.25 cm

volume

10 L
10000 mL

application(s)

cell therapy
gene therapy
life science and biopharma
liquid processing
mAb
ophthalmics
parenterals
pharma/biopharma processes
viral therapy
waste management

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Mobius® Essential Assemblies are standard ready-to-ship bag and tubing assemblies.

Application

Liquid Processing Applications

Features and Benefits

Standard products eliminate design and manufacturing lead times.

Utilizes PureFlex Film and components that can easily scale up to assemblies under Mobius® MyWay program.

Ready-to-download Emprove® Dossier with comprehensive product and regulatory information.

Packaging

5 L - 200 L Bag Assemblies in 5 Pack

Components

PureFlex Film
C-Flex® 374 Tubing 3/8" ID x 5/8" OD, 20" long
C-Flex® 374 Tubing 1/4" ID x 1/2" OD, 20" long
Pinch Clamp 5/8"
Pinch Clamp 3/4"
Boat Port
Hose Barb Non-Valved Coupling Insert 3/8"
Female Luer 1/4" HB
Male Luer

Legal Information

C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service