EFHAB100
EZ-Fit® filtration unit with absorbent pad
pore size 0.45 μm, mixed cellulose esters (MCE) membrane, black filter, volume 100 mL, pkg of Single packaging (EFHAB100I), pkg of Bulk packaging (EFHAB100B), suitable for (suitable for solid and liquid cultivation)
Synonym(s):
Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing
About This Item
Recommended Products
material
black filter
gridded filter
mixed cellulose esters (MCE) membrane
Quality Level
Agency
ISO 7004
sterility
electron beam irradiated
feature
ready-to-use
manufacturer/tradename
EZ-Fit®
packaging
pkg of Bulk packaging (EFHAB100B)
pkg of Single packaging (EFHAB100I)
parameter
100 mL sample volume
capacity
100 mL
filter diam.
47 mm
funnel capacity
100 mL
volume
100 mL
pore size
0.45 μm pore size
input
sample type beverage(s)
sample type pharmaceutical(s)
sample type food(s)
suitability
suitable for (suitable for solid and liquid cultivation)
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with EZ-Fit® System
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
EFHAB100B: Bulk packaging with protective bag (48 units/pack)
Legal Information
configured for
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service