Skip to Content
Merck
All Photos(1)

Key Documents

ECM600

Sigma-Aldrich

uPA Activity Assay Kit

The uPA Activity Assay Kit provides a quick, efficient & sensitive system for evaluation of uPA activity & for screening of uPA inhibitors.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

specific activity

100000 units/mg protein

species reactivity

human

packaging

pkg of ~10 μg standard

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

NCBI accession no.

UniProt accession no.

detection method

colorimetric

shipped in

dry ice

Gene Information

human ... PLAU(5328)

General description

Urokinase-type Plasminogen Activator (uPA) is a 52 kDa serine protease which has been implicated in a number of physiological and pathological processes, including tissue remodeling1, angiogenesis2, fibrinolysis and tumor spread. When bound to its cell surface receptor, uPA is converted from the single chain pro-form uPA to the active 2-chain HMW-uPA. uPA has been shown to play a role in basement membrane degradation, via a cascade involving activation of plasminogen and the matrix metalloproteinases3. Inhibitors of uPA have been shown to slow primary tumor growth and metastasis4-7.

The CHEMICON uPA Activity Assay Kit provides a quick, efficient and sensitive system for evaluation of uPA activity and for screening of uPA inhibitors. The assay is colorimetric and does not require radioactivity or fluorescence equipment. The assay is sensitive over a range of 0.05-50 units of uPA activity.

Test Principle:

The CHEMICON uPA Activity Assay Kit utilizes a chromogenic substrate, which is cleaved by active uPA. Addition of this substrate to a uPA-containing sample results in a colored product, detectable by its Optical Density at 405nm (OD405).

Application:

The CHEMICON uPA Assay Kit is ideal for measurement of uPA activity in purified preparations and cell culture, as well as in serum where pathological conditions such as sepsis exist. The assay is also useful for screening inhibitors of the enzymatic activity of uPA.

Each CHEMICON uPA Activity Assay Kit contains sufficient reagents for the evaluation of 96 samples, including uPA from human urine as a positive control. Duplicate or triplicate samples are suggested.

The CHEMICON uPA Activity Assay Kit is intended for research use only; not for diagnostic or therapeutic applications.

Application

Notes on uPA activity assay sample preparation.



Plasma: Perform venepuncture of the cubial vein, with minimal stasis after a rest period of at least 10min. Discard the first few milliliters of blood. Collect nine volumes of blood in one volume of cold sodium citrate, 0.11 mol/L, in a polycarbonate tube, mix gently and place on ice. Subsequently, prepare platelet-poor plasma by centrifugation in a refrigerated centrifuge (30 min, 2000g). Snap-freeze plasma samples of 0.3 mL to 0.6 mL and store at -60°C. Before use, thaw plasma rapidly in a water bath at 37°C and put on ice.



Tissue: There are numerous publications for the extraction of uPA from tissues. A suitable buffer for extraction of uPA from the membrane fraction requires Triton X100. A non-detergent extract (also called cytosolic fraction) does not require Triton, however the membrane fraction is removed by ultracentrifugation, so uPA activity associated with the membrane fraction is bypassed by measuring only the cytosols. See Peer-Reviewed Papers for some references that describe extraction procedures for uPA.

Cell culture: Isolate cell culture supernatant and use directly.
Research Category
Cell Structure

Components

uPA Positive Control: (Part No. 90058) One lyophilized vial, 1000 units, of uPA from human urine.

Chromogenic Substrate: (Part No. 90057) One 5 mg bottle of Tripeptide with pNA group.

Assay Buffer, 10X: (Part No. 90091) One 5 mL bottle.

Unit Definition

Sensitivity: One unit is defined as the amount of enzyme equal to an international standard tested by the fibrinolytic method of Johnson, et al. {Johnson, A.J., et al. 1969. Thromb. Diath. Haemorrh. 21, 259.}

Storage and Stability

Store kit materials at -20°C for up to 6 months. After reconstitution, store uPA positive control at -70°C and substrate at 2 - 8°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HERG K(+) channel activity is regulated by changes in phosphatidyl inositol 4,5-bisphosphate.
J Bian,J Cui,T V McDonald
Circulation Research null
Mouse NKR-P1B, a novel NK1.1 antigen with inhibitory function.
J R Carlyle,A Martin,A Mehra,L Attisano,F W Tsui,J C Zu?iga-Pflucker
Journal of immunology (Baltimore, Md. : 1950) (1950)
Rev and Rex proteins of human complex retroviruses function with the MMTV Rem-responsive element.
Jennifer A Mertz,Mary M Lozano,Jaquelin P Dudley
Retrovirology null
G Leloup et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 6(10), 1081-1090 (1991-10-01)
The identification of the plasminogen activator (PA) types present in bone and the regulation of their activity by parathyroid hormone (PTH) were investigated in cultures of fetal mouse calvariae with the use of either a chromogenic substrate or a zymographic
Identification of CC chemokine receptor 7 residues important for receptor activation.
Thomas R Ott,Anil Pahuja,Sarah A Nickolls,David G Alleva,R Scott Struthers
The Journal of Biological Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service