Skip to Content
Merck
All Photos(1)

Key Documents

ECM101

Sigma-Aldrich

Human Fibronectin

MILLICOAT® Human Fibronectin Coated Strips (96-Wells), suitable for cell culture

Synonym(s):

Formerly

under the CytoMatrix ™ brand name.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32161000
NACRES:
NA.75

Product Name

Millicoat® Human Fibronectin Coated Strips (96-Wells), Millicoat Cell Adhesion Strips are provided as 12 x 8-well removable strips in a 96-well plate frame for convenience & flexibility in designing assays.

biological source

human

Quality Level

form

lyophilized

species reactivity

human

manufacturer/tradename

Chemicon®
Millicoat®

technique(s)

activity assay: suitable
cell based assay: suitable

input

sample type hematopoietic stem cell(s)
sample type neural stem cell(s)
sample type: human embryonic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type epithelial cells
sample type mesenchymal stem cell(s)
sample type pancreatic stem cell(s)

UniProt accession no.

detection method

colorimetric

shipped in

wet ice

General description

Cell adhesion plays a major role in cellular communication and regulation, and is of fundamental importance in the development and maintenance of tissues. Scientists are continually examining the adhesion and migration of many diverse cell types on various extracellular matrix (ECM) component proteins. Millicoat Cell Adhesion Strips are provided as 12 x 8-well removable strips in a plate frame for convenience and flexibility in designing assays. The wells in rows A - G have been coated with Human Fibronectin. Row H of each strip is coated with BSA which serves as a negative assay control. Cells are seeded onto the coated substrate. Subsequently, adherent cells are fixed and stained. Relative attachment is determined using absorbance readings.

Application

Millicoat Cell Adhesion Strips are provided as 12 x 8-well removable strips in a 96-well plate frame for convenience & flexibility in designing assays.
PROCEDURE:

NOTE: Optimal assay timing and performance may vary for different cell lines but generally can be obtained using subconfluent cell cultures in the assay described below. Subconfluent cultures can be achieved by splitting cells 1 to 2 days prior to performing the assay.

1. Rehydrate the strips with 200 mL of PBS per well for at least 15 minutes at room temperature. Remove the PBS from the rehydrated strips.

2. Prepare a single cell suspension, preferably using a non-enzymatic dissociation buffer. Optimum cell density may be determined by titration of the cells. A common starting range is between 1x10E05 to 1x10E07 cells/mL.

3. Add 100 mL of the diluted cell suspension to each well. Incubate the plate at 37°C for 1 hour in a CO2 incubator. Gently wash the plate 2-3 times with PBS containing Ca2+/Mg2+ (200 mL/well).

4. Add 100 mL of 0.2% crystal violet in 10% ethanol to each well. Incubate for 5 minutes at room temperature. Remove the stain from the wells. Gently wash the plate 3 times with PBS (300 mL/well).

5. Add 100 mL of Solubilization Buffer (A 50/50 mixture of 0.1M NaH2PO4, pH 4.5 and 50% ethanol) to each well. Allow strips to incubate and gently shake at room temperature until the cell-bound stain is completely solubilized; approximately 5 minutes.

6. Determine the absorbance at 540 - 570 nm on a microplate reader.
Research Category
Cell Structure

Packaging

96 wells

Storage and Stability

May be stored at 2-8°C in the foil pouch for at least 3 months. Unused strips may be placed back in the pouch for storage. Ensure that the desiccant remains in the pouch, and that the pouch is securely closed.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLICOAT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interactions between galectin-3 and integrinbeta3 in regulating endometrial cell proliferation and adhesion.
Lei CX, Zhang W, Zhou JP, Liu YK
Human Reproduction null
Regulation of expression and role of leukemia inhibitory factor and interleukin-6 in the uterus of early pregnant pigs.
A Blitek,E Morawska,A J Ziecik
Theriogenology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service