Skip to Content
Merck
All Photos(2)

Key Documents

BRZMBRCFLEX

Millipore

Mobius® Breez 2 mL Microbioreactor

CFlex Tubing, Qty 1

Synonym(s):

Mobius® Breez 2 mL Microbioreactor with CFlex Tubing, Qty 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

sterility

γ-irradiated

technique(s)

cell culture | mammalian: suitable

volume

2 mL

Looking for similar products? Visit Product Comparison Guide

Application

  • Monoclonal antibodies
  • Recombinant proteins

Features and Benefits

A 2 mL Automated Perfusion Cell Culture Platform

For perfusion cell line development, media screening and optimization, and early process development, the ability to run multiple experiments at rapid speed is essential. The Mobius® Breez Microbioreactor System is designed for rapid cell culture experiments, providing an easy-to use solution to produce high quality datasets. It allows for increased productivity and shorter development timelines.

Other Notes

Mobius® Breez Microbioreactor System Services

The pharmaceutical and biotechnology industries are highly regulated. To support you as you navigate this challenging environment, we offer a wide range of services that can help save time and lower costs. All of our services are performed by global experts who have an intimate knowledge of our equipment backed by decades of experience, giving you peace of mind at every step.

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service