BM-116010
Auto2D® IEF Chip pH6-10
used for first-dimension separations on the Auto2D® 2-D Electrophoresis Device., precast gels
Synonym(s):
2D Electrophoresis, 2DE, IEF, IEF Chip, IPG gel, precast gel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
Recommended Products
product line
Auto2D®
General description
IEF Chip is 1st dimensional gel (separation medium) composed of IPG strip gel. IEF Chip pH6-10 is used for first-dimension separations on the Auto2D® 2-D Electrophoresis Device. This gives focused separation on pH6-10 range.
Application
- Differential proteomic analysis
- Post-translational modification
- Protein Isoforms
- Western Blotting
Legal Information
Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service