Skip to Content
Merck
All Photos(1)

Key Documents

AQ160P

Sigma-Aldrich

Rabbit Anti-Mouse IgG Antibody, F(ab′)2, HRP conjugate

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Detect Mouse IgG using this Rabbit anti-Mouse IgG Antibody, F(ab′)2, HRP conjugate validated for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noah J Marcus et al.
Respiratory physiology & neurobiology, 183(2), 67-74 (2012-06-26)
Chronic intermittent hypoxia (CIH) raises arterial pressure, impairs vasodilator responsiveness, and increases circulating angiotensin II (Ang II); however, the role of Ang II in CIH-induced vascular dysfunction is unknown. Rats were exposed to CIH or room air (NORM), and a
Luhui Cai et al.
Molecular medicine reports, 21(2), 959-968 (2020-01-25)
Dental pulp inflammation is a pathological process characterized by local lesions in dental pulp and the accumulation of inflammatory mediators. DNA methylation of cytosine residues is a key epigenetic modification that is essential for gene transcription, and plays pivotal roles
Na Han et al.
Cell biochemistry and function, 39(4), 488-495 (2021-01-13)
Hyaluronic acid (HA) exerts a critical role in the lubricating and buffering properties of synovial fluid in joints. The production of HA is regulated by growth factors, hormones, inflammatory cytokines and mechanical load. The reduction of HA contributes to the
Amanda C Ampey et al.
Molecular and cellular endocrinology, 534, 111368-111368 (2021-06-22)
Uterine artery endothelium undergoes a form of functional adaptation during pregnancy because of an increase in Cx43 communication, resulting in increased Ca2+/IP3 exchange and more synchronous and sustained vasodilator production. We have shown previously that acute exposure to growth factors
Patricia M Garay et al.
Cell reports, 32(6), 108002-108002 (2020-08-14)
Long-lasting forms of synaptic plasticity such as synaptic scaling are critically dependent on transcription. Activity-dependent transcriptional dynamics in neurons, however, remain incompletely characterized because most previous efforts relied on measurement of steady-state mRNAs. Here, we use nascent RNA sequencing to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service