Skip to Content
Merck
All Photos(1)

Key Documents

AQ132P

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, F(ab′)2, HRP conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Detect Rabbit IgG using this Goat anti-Rabbit IgG Antibody, F(ab′)2, HRP conjugate validated for use in ELISA & WB.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Liu et al.
International journal of molecular medicine, 46(5), 1671-1682 (2020-10-02)
Oxidized low‑density lipoprotein (ox‑LDL)‑induced endothelial cell (EC) injury is a risk factor for atherosclerosis. Therefore, the present study aimed to investigate the effects of insulin‑receptor substrate 1 (IRS‑1) on injury to ox‑LDL‑exposed ECs. For this purpose, thoracic aorta tissues were
Lawrence Petherbridge et al.
Vaccines, 11(11) (2023-11-25)
Respiratory syncytial virus (RSV) infection and shingles are two viral diseases that affect older adults, and a combined vaccine to protect against both could be beneficial. RSV infection causes hospitalisations and significant morbidity in both children and adults and can
Ying-Tao Lv et al.
Adipocyte, 10(1), 28-37 (2021-01-05)
Obesity is an important public-health problem worldwide. This study aimed to determine effects of porphyromonas gingivalis lipopolysaccharide (Pg-LPS) on adipocytes injuries and explore associated mechanisms. Adipocytes were isolated from SD rats. pLVX-XBP1 (XBP1 over-expression) and pLVX-XBP1-RNAi (silencing XBP1) were structured
Gene and protein expression and cellular localisation of cytochrome P450 enzymes of the 1A, 2A, 2C, 2D and 2E subfamilies in equine intestine and liver.
Tyden, E; Tjalve, H; Larsson, P
Acta Veterinaria Scandinavica null
Xiao-Fei Zhang et al.
Molecular carcinogenesis, 55(1), 64-76 (2015-01-06)
Ubiquitination factor E4B (UBE4B) has been speculated to have contradictory functions upon tumorigenesis as an oncogene or tumor suppressor in different types of cancers. We investigated the expression and prognostic role of UBE4B in primary hepatocellular carcinoma (HCC) using cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service