Skip to Content
Merck
All Photos(1)

Key Documents

AP503P

Sigma-Aldrich

Goat Anti-Mouse IgG γ chain Antibody, HRP conjugate, Species Adsorbed

Chemicon®, from goat

Synonym(s):

Goat Anti-Mouse IgG HRP, HRP-Conjugated Goat Anti-Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

IgG is one of the most abundant protein in human serum. IgG is important for our defence against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Reacts with heavy chain of mouse IgG1, IgG2a, IgG2b and IgG3. Absorbed for mouse IgM and IgA, pooled human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins.
The antibody reacts with the heavy chain of mouse IgG1, IgG2a, IgG2b and IgG3 as demonstrated by ELISA and flow cytometry. Minimal cross reactivity with human immunoglobulins.

Immunogen

Epitope: IgG γ chain
Prepared from purified mouse IgG γ chain. Pooled antisera from goats hyperimmunized with mouse IgG paraproteins.

Application

ELISA: 1:4,000-1:8,000

Optimal working dilutions must be determined by the end user.
Goat anti-Mouse IgG γ chain Antibody, HRP conjugate, Species Adsorbed is an antibody against Mouse IgG γ chain for use in ELISA.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Fragment Specific Secondary Antibodies

Physical form

Affinity Chromatography
Purified by affinity chromatography on pooled mouse IgG covalently linked to agarose. Liquid in 50% glycerol/50% PBS, pH 7.4.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. For long term storage, aliquot and store at -20°C. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harrison Dulin et al.
Journal of virology, 98(2), e0157123-e0157123 (2024-01-11)
In pandemic scenarios involving novel human pathogenic viruses, it is highly desirable that vaccines induce strong neutralizing antibodies as quickly as possible. However, current vaccine strategies require multiple immunization doses to produce high titers of neutralizing antibodies and are poorly
A J Conley et al.
Theriogenology, 108, 192-200 (2017-12-12)
Studies in mares have examined serum inhibin concentrations using immuno-assays unable to distinguish dimeric inhibin-A from inhibin-B isoforms. Inhibin-A and inhibin-B immuno-assays were used to investigate concentrations in cyclic mares, young and old (6 vs 19 years old, respectively) mares
Robert Davis et al.
Molecular microbiology, 107(1), 116-131 (2017-11-07)
Transcriptional regulation is the key to ensuring that proteins are expressed at the proper time and the proper amount. In Escherichia coli, the transcription factor cAMP receptor protein (CRP) is responsible for much of this regulation. Questions remain, however, regarding
Xi Wang et al.
Oncology reports, 44(3), 1025-1036 (2020-07-25)
Emerging evidence has demonstrated that abnormally expressed long non‑coding (lnc) RNAs contribute to drug resistance in various types of malignancy. LncRNA antisense non‑coding RNA in the inhibitor of cyclin‑dependent kinase 4 locus (ANRIL) exerts oncogenic activity and acts as a
Meito Shibuya et al.
iScience, 24(10), 103131-103131 (2021-10-09)
Current influenza vaccines do not typically confer cross-protection against antigenically mismatched strains. To develop vaccines conferring broader cross-protection, recent evidence indicates the crucial role of both cross-reactive antibodies and viral-specific CD4+ T cells; however, the precise mechanism of cross-protection is unclear. Furthermore, adjuvants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service