Skip to Content
Merck
All Photos(1)

Key Documents

AP184C

Sigma-Aldrich

Donkey Anti-Sheep IgG Antibody, Cy3 conjugate, Species Adsorbed

Chemicon®, from donkey

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

sheep

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Donkey anti-Sheep IgG Antibody, Cy3 conjugate, Species Adsorbed is an antibody against Donkey Sheep IgG for use in IF.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salma Pathan-Chhatbar et al.
The Journal of biological chemistry, 293(3), 863-875 (2017-12-02)
The metazoan actin cytoskeleton supports a wide range of contractile and transport processes. Recent studies have shown how the dynamic association with specific tropomyosin isoforms generates actin filament populations with distinct functional properties. However, critical details of the associated molecular
Sahar Mani et al.
Clinical and experimental pharmacology & physiology, 50(9), 728-737 (2023-06-19)
Parkinson's disease (PD) is a neurodegenerative disorder characterized by a progressive loss of dopaminergic neurons in the substantia nigra, for which no disease-modifying treatments are available yet. Thus, developing new neuroprotective drugs with the potential to delay or stop the
Johannes Roeles et al.
Nature communications, 10(1), 4651-4651 (2019-10-13)
Mammalian oocytes assemble a bipolar acentriolar microtubule spindle to segregate chromosomes during asymmetric division. There is increasing evidence that actin in the spindle interior not only participates in spindle migration and positioning but also protects oocytes from chromosome segregation errors
Jianying Zhang et al.
Cell and tissue research, 384(2), 367-387 (2021-01-27)
This study aimed to characterize porcine Achilles tendon (PAT) in terms of its structural components, vascularity, and resident tendon cells. We found that PAT is composed of a paratenon sheath, a core of fascicles, and an endotenon/interfascicular matrix (IFM) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service