Skip to Content
Merck
All Photos(1)

Documents

AP106

Sigma-Aldrich

Rabbit Anti-Goat IgG Antibody

2.0 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

concentration

2.0 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with Goat IgG gamma chain, as well as the light chains from all Goat immunoglobulin classes. Monospecific on IEP against whole Goat Serum. Single band on IEP at a concentration of 10 mg/mL when run against anti-whole serum.

Immunogen

Highly purified Goat IgG isolated from pooled normal Goat Serum.

Application

Detect Goat IgG using this Rabbit anti-Goat IgG Antibody validated for use in ELISA, IP & WB.
NOTE:

Due to the wide range of variable immunoassays for which this product is intended, it is necessary that each laboratory establish individual performance characteristics for all assay components in any specific assay system.

For use in Research and Manufacturing only.

Not intended for direct use in diagnostic applications.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Nonsterile liquid. Buffer=0.02M Phosphate Buffer,

0.25M Sodium Chloride and 0.1% Sodium Azide. Filtered through a 0.45 micron membrane filter.

Storage and Stability

Store at +2-8°C for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun-Chun Chiang et al.
Anticancer research, 37(11), 6215-6221 (2017-10-25)
Pancreatic neuroendocrine tumors (PanNETs) are usually diagnosed in an advanced stage. Most patients with PanNETs die of metastasis. Vascular endothelial growth factor-A (VEGF-A) is a strong stimulator of angiogenesis and tumor metastasis. We aimed to investigate the effect of MART-10
Up-regulation of METCAM/MUC18 promotes motility, invasion, and tumorigenesis of human breast cancer cells.
Zeng, GF; Cai, SX; Wu, GJ
BMC Cancer null
Nobuko Harada et al.
Surgery, 134(3), 480-491 (2003-10-14)
In ischemia/reperfusion (I/R) injury, a massive generation of reactive oxygen species (ROS) after reperfusion is a critical factor. Rac, a member of the Rho GTPase superfamily, plays important roles in the production of ROS and activation of nuclear factor-kappaB (NF-kappaB)
Enforced expression of METCAMMUC18 increases tumorigenesis of human prostate cancer lNCaP cells in nude Mice.
Wu GJ, Wu MW, Wang C, Liu Y
The Journal of urology null
Yan Zhou et al.
Journal of neurochemistry, 102(3), 667-678 (2007-04-04)
Microglial cells release monocyte chemoattractant protein-1 (MCP-1) which amplifies the inflammation process by promoting recruitment of macrophages and microglia to inflammatory sites in several neurological diseases. In the present study, dexamethasone (Dex), an anti-inflammatory and immunosuppressive drug has been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service