Skip to Content
Merck
All Photos(3)

Key Documents

AB9748-I

Sigma-Aldrich

Anti-14-3-3 Antibody (pan)

from rabbit, purified by affinity chromatography

Synonym(s):

14-3-3 protein beta/alpha, KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1, 14-3-3 protein epsilon, 14-3-3E, 14-3-3 protein eta, Protein AS1, 14-3-3 protein gamma, 14-3-3 protein theta, 14-3-3 protein T-cell, 14-3-3 protein tau, Protein HS1, 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), mouse (based on 100% sequence homology), Xenopus (based on 100% sequence homology), fish (based on 100% sequence homology), zebrafish (based on 100% sequence homology), rabbit (based on 100% sequence homology), chicken (based on 100% sequence homology), sheep (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

ambient

target post-translational modification

unmodified

General description

14-3-3 protein beta/alpha (KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1), epsilon (14-3-3E), eta (Protein AS1), gamma (KCIP-1, Protein kinase C inhibitor protein 1), theta (14-3-3 protein T-cell, 14-3-3 protein tau, Protein HS1), zeta/delta (KCIP-1, Protein kinase C inhibitor protein 1) are encoded by the YWHAB, YWHAE, YWHAH, YWHAG, YWHAQ, YWHAZ gene (Gene ID 7529, 7531, 7533, 7532, 10971, 7534, respectively) in human. Mammals express seven distinct 14-3-3 isoforms (gamma, epsilon, beta, zeta, sigma, theta, tau) that form multiple homo- and hetero- dimmers, thus rendering 14-3-3 as a key multifunctional regulatory molecule. 14-3-3 isomers have been implicated in several neurological disorders: for example isoforms gamma, epsilon and beta are expressed in the cerebrospinal fluid of Creutzfeldt-Jakob patients, but not patients suffering from other neurological conditions such as Alzheimer′s. The 14-3-3 pan antibody is one of a panel of antibodies that were generated against 14-3-3 isoforms and have demonstrated their potential for use as an ELISA-based diagnostic tool. These antibodies will allow the research to investigate the role of this protein in key cellular processes and neurological disorders such as Creutzfeldt-Jakob, Alzheimer′s and Parkinson′s disease.

Specificity

This antibody targets the N-terminal end sequences of 14-3-3 alpha/beta (YWHAB), gamma (YWHAG), zeta/delta (YWHAZ), epsilon (YWHAE), eta (YWHAH or YWHA1), and theta (YWHAQ).

Immunogen

A mixture of KLH-conjugated linear peptides corresponding the the N-terminal end sequences of human 14-3-3 alpha/beta (YWHAB), gamma (YWHAG), zeta/delta (YWHAZ), epsilon (YWHAE), eta (YWHAH or YWHA1), and theta (YWHAQ).

Application

Anti-14-3-3 pan, Cat. No. AB9748-I, is a highly specific rabbit polyclonal antibody, that targets 14-3-3 protein and has been tested in Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting and .
Immunohistochemistry Analysis: A 1:5,000 dilution from a representative lot detected 14-3-3 immunoreactivity in human prostate cancer, cerebellum, and tonsil tissue sections.

Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipitated 14-3-3 from 500 µg of A431 cell lysate.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected 14-3-3 in 10 µg of A431 cell lysate.

Target description

~28 kDa observed. 28.08/27.85 kDa (human isoform Long/Short; YWHAB), 27.75 kDa (human YWHAZ), 29.17 kDa (human YWHAE), 28.22 kDa (human YWHAH or YWHA1), 28.30 kDa (human YWHAG), 27.76 kDa (human YWHAQ) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: AB9748

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kolonko-Adamska et al.
Frontiers in molecular biosciences, 10, 1215550-1215550 (2023-09-01)
Methoprene-tolerant (Met) and germ cell-expressed (Gce) proteins were shown to be juvenile hormone (JH) receptors of Drosophila melanogaster with partially redundant functions. We raised the question of where the functional differentiation of paralogs comes from. Therefore, we tested Met and
François Enchéry et al.
Communications biology, 4(1), 1292-1292 (2021-11-18)
Nipah virus (NiV) is a highly pathogenic emerging bat-borne Henipavirus that has caused numerous outbreaks with public health concerns. It is able to inhibit the host innate immune response. Since the NF-κB pathway plays a crucial role in the innate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service