Skip to Content
Merck
All Photos(1)

Key Documents

AB1513P

Sigma-Aldrich

Anti-Brain Derived Neurotrophic Factor Antibody

Chemicon®, from sheep

Synonym(s):

BDNF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

sheep

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, chicken

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

Specificity

Recognizes Brain Derived Neurotrophic Factor (BDNF). Neutralizes BDNF, but not other neurotrophins. By one site ELISA, less than 1% cross-reactivity against mouse NGF, recombinant human NT3 or neurotrophin 4. The antibody was tested on human and rat derived samples, but based on sequence similarity it is expected to react with mouse and other mammalian species. The peptide sequences of human, rat and mouse BDNF are identical.

Immunogen

Recombinant Human Brain Derived Neurotrophic Factor (BDNF).

Application

Anti-Brain Derived Neurotrophic Factor Antibody detects level of Brain Derived Neurotrophic Factor & has been published & validated for use in ELISA, IH, FUNC & WB.
Immunohistochemistry: 1-5 μg/mL (see suggested protocol).

Immunoblotting: 1-5 μg/mL we recommend preparing the BDNF samples for western blot by the method of Semba-Katoh, R et.al (J. Neurochem. 69(1):34-42, 1997) because BDNF is not easily extracted in standard buffers. Dissected tissues should be homogenized in 10 volumes of 100mM phosphate buffer containing 1mM EDTA, 2M guanidine hydrochloride (pH7.2), and three protease inhibitors, 10mM N-ethylmaleimide, 0.36mM pepstatin, and 1mM PMSF. Homogenates are then sonicated and centrifuged at 46K x g for 30 minutes at 4°C. BDNF migrates as ~12-18kDa monomer, with a 28-30kDa homodimer in most samples.

ELISA: 1-5 μg/mL

Inhibition of biological activity in vitro: Complete inhibition of BDNF activity on embryonic chick sensory neurons occurs at 1-5 μg/mL. Biological inhibition protocols can be found in Finn et al. (J. Neurocytol. 15:169-176, 1986).
BIOLOGICAL ACTIVITY: Neutralizes BDNF, but not other neurotrophins.

Use neat for in vivo studies.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain

Target description

~12-18kDa monomer, with a 28-30kDa homodimer in most samples.

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin. Lyophilized from PBS, no preservatives. Reconstitute with 500 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Glycerol (1:1) can be added for additional stability. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Deyama et al.
Translational psychiatry, 12(1), 178-178 (2022-05-17)
Ketamine, an N-methyl-D-aspartate receptor antagonist, exerts rapid and sustained antidepressant actions. Preclinical studies demonstrated that the release of brain-derived neurotrophic factor (BDNF) and vascular endothelial growth factor in the medial prefrontal cortex (mPFC) is essential for the antidepressant-like effects of
Juan Gabriel Riboldi et al.
Communications biology, 7(1), 1032-1032 (2024-08-23)
Glutamate is involved in fundamental functions, including neuronal plasticity and memory. Astrocytes are integral elements involved in synaptic function, and the GLT-1 transporter possesses a critical role in glutamate uptake. Here, we study the role of GLT-1, specifically located in
Mariana Alonso et al.
Learning & memory (Cold Spring Harbor, N.Y.), 12(5), 504-510 (2005-10-06)
Information storage in the brain is a temporally graded process involving different memory phases as well as different structures in the mammalian brain. Cortical plasticity seems to be essential to store stable long-term memories, although little information is available at
Ting-Chun Lin et al.
Frontiers in cellular neuroscience, 17, 1170251-1170251 (2023-05-30)
Intracerebral hemorrhage (ICH) enhances neurogenesis in the subventricular zone (SVZ); however, the mechanism is not fully understood. We investigated the role of brain-derived neurotrophic factor (BDNF) in post-ICH neurogenesis in a rodent model and in patients with ICH using cerebrospinal
S Rasika et al.
Neuron, 22(1), 53-62 (1999-02-23)
New neurons are incorporated into the high vocal center (HVC), a nucleus of the adult canary (Serinus canaria) brain that plays a critical role in the acquisition and production of learned song. Recruitment of new neurons in the HVC is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service